Skip to Content
Merck
All Photos(2)

Key Documents

289841

Sigma-Aldrich

Calcium oxalate hydrate

Synonym(s):

Calcium ethanedioate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2CaO4 · xH2O
CAS Number:
Molecular Weight:
128.10 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

form

powder

concentration

57.0-62.0% (by KMnO4, %C2O4, titration)

SMILES string

O.O=C1O[Ca]OC1=O

InChI

1S/C2H2O4.Ca.H2O/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);;1H2/q;+2;/p-2

InChI key

LQHWSGSWNOHVHO-UHFFFAOYSA-L

Related Categories

Application

  • Local concentration controls the hydrate phase of calcium oxalate.: This research demonstrates how adjusting the titration rate of calcium solution and stirring speed can influence the formation of different hydrate phases of calcium oxalate. Such control is critical for applications requiring specific crystal forms, such as in bioceramics and industrial processes (Wang et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Sujatha et al.
International braz j urol : official journal of the Brazilian Society of Urology, 41(3), 511-520 (2015-07-23)
Urolithiasis is a common urological disorder responsible for serious human affliction and cost to the society with a high recurrence rate. The aim of the present study was to systematically evaluate the phlorotannin rich extract of Sargassum wightii using suitable
Xin Wang et al.
International journal of molecular medicine, 45(2), 375-384 (2020-01-03)
MicroRNAs (miRNAs or miRs) are critical regulators in various diseases. In the current study, the role of miR‑30c‑5p in the formation of sodium oxalate‑induced kidney stones was investigated. For this purpose, human renal tubular epithelial cells (HK‑2 cells) were incubated
J Tonannavar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 154, 20-26 (2015-10-27)
We present in this paper accurate and reliable Raman and IR spectral identification of mineral constituents in nine samples of renal calculi (kidney stones) removed from patients suffering from nephrolithiasis. The identified mineral components include Calcium Oxalate Monohydrate (COM, whewellite)
A Schmalenberger et al.
Scientific reports, 5, 12187-12187 (2015-07-23)
Trees and their associated rhizosphere organisms play a major role in mineral weathering driving calcium fluxes from the continents to the oceans that ultimately control long-term atmospheric CO2 and climate through the geochemical carbon cycle. Photosynthate allocation to tree roots
Daniel Bravo et al.
Archives of microbiology, 197(1), 65-77 (2014-11-10)
The oxalate-carbonate pathway (OCP) is a biogeochemical set of reactions that involves the conversion of atmospheric CO2 fixed by plants into biomass and, after the biological recycling of calcium oxalate by fungi and bacteria, into calcium carbonate in terrestrial environments.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service