Skip to Content
Merck
All Photos(1)

Key Documents

289183

Sigma-Aldrich

Neodymium(III) chloride hexahydrate

99.9% trace metals basis

Synonym(s):

Neodymium trichloride hexahydrate, trichloroneodymium hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NdCl3 · 6H2O
CAS Number:
Molecular Weight:
358.69
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

crystals and lumps

reaction suitability

reagent type: catalyst
core: neodymium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

mp

124 °C (lit.)

SMILES string

O.O.O.O.O.O.Cl[Nd](Cl)Cl

InChI

1S/3ClH.Nd.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

PCORICZHGNSNIZ-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Neodymium(III) chloride hexahydrate may be used as a source of Nd3+ ions, to be used to dope AgBr crystals, known to exhibit lasing effects.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luminescence method for the study of Nd3+ ions diffusion in AgBr crystals.
Nagli L, et al.
Journal of Applied Physics, 85(4), 2114-2118 (1999)
Emilia Palo et al.
Langmuir : the ACS journal of surfaces and colloids, 34(26), 7759-7766 (2018-06-15)
Aqueous solutions are the basis for most biomedical assays, but they quench the upconversion luminescence significantly. Surface modifications of upconverting nanoparticles are vital for shielding the obtained luminescence. Modifications also provide new possibilities for further use by introducing attaching sites

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service