Skip to Content
Merck
All Photos(1)

Key Documents

281867

Sigma-Aldrich

Diethyl acetamidomalonate-2-13C

≥99 atom % 13C, ≥99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONH13CH(CO2C2H5)2
CAS Number:
Molecular Weight:
218.21
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

≥99 atom % 13C

Quality Level

Assay

≥99% (CP)

form

solid

bp

185 °C/20 mmHg (lit.)

mp

95-98 °C (lit.)

mass shift

M+1

SMILES string

CCOC(=O)[13CH](NC(C)=O)C(=O)OCC

InChI

1S/C9H15NO5/c1-4-14-8(12)7(10-6(3)11)9(13)15-5-2/h7H,4-5H2,1-3H3,(H,10,11)/i7+1

InChI key

ISOLMABRZPQKOV-CDYZYAPPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sambasivarao Kotha et al.
Amino acids, 41(4), 933-936 (2010-11-10)
Strategically a new approach for the synthesis tetrahydro-β-carboline unit with the aid of diethyl acetamidomalonate as a glycine equivalent has been described.
Soraia Veloso Silva Santana et al.
Journal of esthetic and restorative dentistry : official publication of the American Academy of Esthetic Dentistry ... [et al.], 21(6), 397-404 (2009-12-17)
Surface sealants may reduce or avoid problems related to the marginal interface. The aim of this study was to evaluate the microleakage in resin composite Class V restorations sealed with an adhesive system (Xeno III [Dentsply, Konstanz, Germany]), a sealant
Zsolt Rapi et al.
Carbohydrate research, 365, 61-68 (2012-12-12)
The synthesis of four new ribo-hexopyranoside-based chiral lariat ethers of monoaza-15-crown-5 type and two altropyranoside-based crown ethers were elaborated. Our syntheses utilized the regioselective ring opening of the oxiran moiety of the 2,3-anhydro sugars by nucleophilic reagents to afford the
G K Powell et al.
Preparative biochemistry, 11(3), 339-350 (1981-01-01)
This paper describes the complete chemical synthesis of 4-methylene-DL-glutamic acid from diethylmalonate, formaldehyde and diethyl acetamidomalonate. The amino acid was obtained pure following ion-exchange chromatography and/or crystallization from hot water in an overall yield of 30% based on the amount

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service