Merck
  • Synthesis of ribo-hexopyranoside- and altrose-based azacrown ethers and their application in an asymmetric Michael addition.
Synthesis of ribo-hexopyranoside- and altrose-based azacrown ethers and their application in an asymmetric Michael addition.
Carbohydrate research (2012-12-12)
Zsolt Rapi, Péter Bakó, György Keglevich, Áron Szöllősy, László Drahos, László Hegedűs
ABSTRACT
The synthesis of four new ribo-hexopyranoside-based chiral lariat ethers of monoaza-15-crown-5 type and two altropyranoside-based crown ethers were elaborated. Our syntheses utilized the regioselective ring opening of the oxiran moiety of the 2,3-anhydro sugars by nucleophilic reagents to afford the key intermediates. The reaction of methyl-2,3-anhydro-4,6-O-benzylidene-α-D-mannopyranoside with ethanolamine is especially of interest to afford a 3-substituted altropyranoside. One of the ribo-hexopyranoside-based lariat ethers with a 4-methoxyphenyl substituent induced an enantioselectivity of 80% when used as catalyst in the Michael addition of diethyl acetamidomalonate to trans-β-nitrostyrene under phase transfer catalytic conditions.
Sigma-Aldrich
Diethyl acetamidomalonate-2-13C, ≥99 atom % 13C, ≥99% (CP)
Sigma-Aldrich
Diethyl acetamidomalonate, 98%
Sigma-Aldrich
trans-β-Nitrostyrene, 99%
Sigma-Aldrich
15-Crown-5, 98%