Skip to Content
Merck
All Photos(1)

Key Documents

277681

Sigma-Aldrich

3-Methyl-3-oxetanemethanol

98%

Synonym(s):

3-Hydroxymethyl-3-methyl-oxetane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O2
CAS Number:
Molecular Weight:
102.13
Beilstein:
102773
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.446 (lit.)

bp

80 °C/40 mmHg (lit.)

density

1.024 g/mL at 25 °C (lit.)

functional group

ether
hydroxyl

storage temp.

2-8°C

SMILES string

CC1(CO)COC1

InChI

1S/C5H10O2/c1-5(2-6)3-7-4-5/h6H,2-4H2,1H3

InChI key

NLQMSBJFLQPLIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Methyl-3-oxetanemethanol has been used in the preparation of:
  • star-shaped copolymer consisting of a hyperbranched poly(3-methyl-3-oxetanemethanol) core and polytetrahydrofuran arms
  • pyridyl disulfide-functionalized cyclic carbonate monomer, required for the synthesis of of functional poly(ε-caprolactone) containing pendant pyridyl disulphide groups
  • δ-lactams

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of a Star-Shaped Copolymer with a Hyperbranched Poly (3-methyl-3-oxetanemethanol) Core and Tetrahydrofuran Arms by One-Pot Copolymerization.
Hou J and Yan D.
Macromolecular Rapid Communications, 23(8), 456-459 (2002)
Functional poly (e-caprolactone) s via copolymerization of e-caprolactone and pyridyl disulfide-containing cyclic carbonate: controlled synthesis and facile access to reduction-sensitive biodegradable graft copolymer micelles.
Chen W, et al.
Macromolecules, 46(3), 699-707 (2013)
Bhooma Raghavan et al.
The Journal of organic chemistry, 71(5), 2151-2154 (2006-02-25)
A concise, stereoselective synthesis of alpha-substituted gamma-lactams is reported. gamma-Lactam scaffolds 2 and 3, possessing an Evans' chiral auxiliary and two types of N substituents, were successfully alkylated with different electrophiles to give alpha-substituted gamma-lactams with reasonable diastereoselectivities. The use
Gintare Krucaite et al.
Molecules (Basel, Switzerland), 26(7) (2021-04-04)
A group of polyethers containing electroactive pendent 4,7-diarylfluorene chromophores have been prepared by the multi-step synthetic route. Full characterization of their structures has been presented. The polymeric materials represent derivatives of high thermal stability with initial thermal degradation temperatures in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service