Skip to Content
Merck
All Photos(4)

Key Documents

302309

Sigma-Aldrich

2-Cyanoethyl N,N-diisopropylchlorophosphoramidite

Cl 13.5-15.5 %

Synonym(s):

Chloro(diisopropylamino)-β-cyanoethoxyphosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2NP(Cl)OCH2CH2CN
CAS Number:
Molecular Weight:
236.68
Beilstein:
3588525
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

composition

Cl, 13.5-15.5%

refractive index

n/D 1.476-1.480

bp

103 -105 °C/0.1 hPa

density

1.061 g/mL at 25 °C (lit.)

functional group

amine
nitrile

storage temp.

−20°C

SMILES string

CC(C)N(C(C)C)P(Cl)OCCC#N

InChI

1S/C9H18ClN2OP/c1-8(2)12(9(3)4)14(10)13-7-5-6-11/h8-9H,5,7H2,1-4H3

InChI key

QWTBDIBOOIAZEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Cyanoethyl N,N-diisopropylchlorophosphoramidite was employed:
  • for selective monophosphorylation of carbohydrates and nucleosides
  • for conversion of protected ribonucleosides to phosphoramidites
  • as phosphitylating reagent for 3′-hydroxyl groups in the synthesis of oligodeoxyribonucleotides
  • in a scalable, solution-phase oligonucleotide synthesis employing phosphoramidite chemistry and DMT-, iBu- and Bz-protected monomers. Intermediates were isolated by extraction without further purification

related product

Product No.
Description
Pricing

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yousef Ahmadibeni et al.
The Journal of organic chemistry, 70(3), 1100-1103 (2005-01-29)
Two classes of aminomethyl polystyrene resin-bound linkers of p-acetoxybenzyl alcohol were subjected to reactions with 2-cyanoethyl N,N-diisopropylchlorophosphoramidite to produce the corresponding polymer-bound phosphitylating reagents. These were reacted with a number of unprotected nucleosides and carbohydrates in the presence of 1H-tetrazole.
Tetrahedron Letters, 34, 6189-6189 (1993)
Tetrahedron Letters, 45, 509-509 (2004)
A Ono et al.
Bioconjugate chemistry, 4(6), 499-508 (1993-11-01)
Novel 2'-deoxyuridine analogues carrying aminoalkyl linkers at the 1'-position of the sugar residues were synthesized and incorporated into oligonucleotides, then intercalating groups such as an anthraquinone derivative and a pyrene derivative were attached to the amino groups. Duplexes consisting of
Organic Process Research & Development, 10, 1238-1238 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service