Skip to Content
Merck
All Photos(1)

Key Documents

262099

Sigma-Aldrich

Phosphorus(V) oxychloride

99.999%

Synonym(s):

Phosphorus(V) oxide chloride, Phosphoryl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
POCl3
CAS Number:
Molecular Weight:
153.33
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22
bp:
105.8 °C (lit.)
vapor pressure:
104 mmHg ( 50 °C)
28 mmHg ( 20 °C)

vapor density

5.3 (vs air)

vapor pressure

104 mmHg ( 50 °C)
28 mmHg ( 20 °C)

Assay

99.999%

form

liquid

pH

1 (20 °C)

bp

105.8 °C (lit.)

mp

1.25 °C (lit.)

density

1.645 g/mL at 25 °C (lit.)

functional group

phosphine oxide

SMILES string

ClP(Cl)(Cl)=O

InChI

1S/Cl3OP/c1-5(2,3)4

InChI key

XHXFXVLFKHQFAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phosphorus(V) oxychloride can be used as:
  • A dehydrating agent in organic synthesis.
  • A reagent in the cyclization of substituted aryl amides by Bischler−Napieralski cyclization reaction.
  • A precursor for phosphorus diffusion in the p-type silicon solar cells processing.

It can also be used in the Vilsmeier-Haack reaction to synthesize aromatic aldehydes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric transfer hydrogenation of imines catalyzed by a Noyori-type Ru (II) complex-a parametric study
Pechavcek J, et al.
Tetrahedron Asymmetry, 24(4), 233-239 (2013)
Isocyanide 2.0
Patil P, et al.
Green Chemistry, 22(20), 6902-6911 (2020)
Optimization of phosphorus emitter formation from POCl 3 diffusion for p-type silicon solar cells processing
Ghembaza H, et al.
Silicon, 10(2), 377-386 (2018)
Rapid one-pot radiosynthesis of [carbonyl-. sup. 11C] formamides from primary amines and [. sup. 11C] CO. sub. 2.
Luzi Federico, et al.
EJNMMI radiopharmacy and chemistry, 5(1), 377-386 (2020)
Bo Qin et al.
Chemistry, an Asian journal, 6(12), 3298-3305 (2011-07-23)
POCl(3)-mediated one-pot macrocyclization allows the highly selective formation of five-residue macrocycles that are rigidified by internally placed intramolecular hydrogen bonds. Mechanistic investigation by using tailored competition experiments and kinetic simulation provides a comprehensive model, supporting a chain-growth mechanism underlying the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service