Skip to Content
Merck
All Photos(1)

Key Documents

261556

Sigma-Aldrich

(L)-Dehydroascorbic acid

Synonym(s):

DHAA, Dehydro-L-ascorbate, Dehydro-L-ascorbic acid, L-threo-Dehydroascorbic acid, oxidized ascorbic acid, oxidized vitamin C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O6
CAS Number:
Molecular Weight:
174.11
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.22

form

powder

mp

228 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

OC[C@H](O)[C@H]1OC(=O)C(=O)C1=O

InChI

1S/C6H6O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-8H,1H2/t2-,5+/m0/s1

InChI key

SBJKKFFYIZUCET-JLAZNSOCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(L)-Dehydroascorbic acid (DHA) is a potent glycation agent that is produced by the oxidation of ascorbic acid (vitamin C). In vitro studies revealed that DHA is significantly more reactive than glucose and fructose in the glycation of lens proteins. Thus, DHA could be a critical precursor for the in vivo formation of advanced glycation end products (AGEs).
DHA is a dimer when it is solid, but when it is dissolved in a solution, it becomes a monomer.

Application

(L)-Dehydroascorbic acid can be used as a starting material to synthesize:
  • Pyrano[3,4-b]indole derivatives by reacting with 2-hydroxymethylindole.
  • (−)-ascorbyl phloroglucinol via stereoselective C-C bond formation reaction with phloroglucinol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Els Vossen et al.
Meat science, 91(1), 29-35 (2012-01-10)
The effect of sodium ascorbate (SA; 500, 750, 1000 mg/kg) and sodium nitrite (SN; 40, 80, 120 mg/kg) doses on the shelf-life stability of liver pâtés was investigated in a full factorial design. Clear dose-dependent responses of the added SN
Oliver Reihl et al.
Carbohydrate research, 339(3), 483-491 (2004-03-12)
Covalently cross-linked proteins are among the major modifications caused by the advanced Maillard reaction. So far, the chemical nature of these aggregates is largely unknown. L-dehydroascorbic acid (DHA, 5), the oxidation product of L-ascorbic acid (vitamin C), is known as
Jens Lykkesfeldt
Nutrition research (New York, N.Y.), 32(1), 66-69 (2012-01-21)
Ascorbate and dehydroascorbic acid are frequently used as biomarkers of oxidative stress, but their lack of stability ex vivo and rapid postsampling interconversion continue to result in erroneous reference values. One problem is the large variety of vacutainer devices used
A biomimetic synthesis of (-)-ascorbyl phloroglucinol and studies toward the construction of ascorbyl-modified catechin natural products and analogues
Belapure SA, et al.
Tetrahedron, 67(48), 9265-9272 (2011)
A Wesley Burks et al.
Pediatric allergy and immunology : official publication of the European Society of Pediatric Allergy and Immunology, 26(4), 316-322 (2015-04-08)
Children with cow's milk allergy (CMA) are at risk for inadequate nutritional intake and growth. Dietary management of CMA, therefore, requires diets that are not only hypoallergenic but also support adequate growth in this population. This study assessed growth of

Articles

Simultaneous analysis of ascorbic acid and dehydroascorbic acid in various food samples by High Performance Thin Layer Chromatography (HPTLC).

Simultaneous analysis of ascorbic acid and dehydroascorbic acid in various food samples by High Performance Thin Layer Chromatography (HPTLC).

Simultaneous analysis of ascorbic acid and dehydroascorbic acid in various food samples by High Performance Thin Layer Chromatography (HPTLC).

Simultaneous analysis of ascorbic acid and dehydroascorbic acid in various food samples by High Performance Thin Layer Chromatography (HPTLC).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service