Skip to Content
Merck
All Photos(1)

Key Documents

24940

Sigma-Aldrich

4-Chloro-3-methylphenol

≥98.0% (HPLC)

Synonym(s):

4-Chloro-m-cresol, PCMC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CH3)OH
CAS Number:
Molecular Weight:
142.58
Beilstein:
1237629
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

form

pellets

bp

235 °C (lit.)

mp

63-65 °C (lit.)

solubility

methanol: soluble 1 g/10 mL, clear, colorless
water: soluble

SMILES string

Cc1cc(O)ccc1Cl

InChI

1S/C7H7ClO/c1-5-4-6(9)2-3-7(5)8/h2-4,9H,1H3

InChI key

CFKMVGJGLGKFKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chloro-3-methylphenol is an organic compound belonging to the class of monochlorinated m-cresol. It is used as an organic building block in chemical synthesis.

Application

4-Chloro-3-methylphenol has been used in paint formulations.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

244.4 °F - closed cup

Flash Point(C)

118.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roel Mallants et al.
The Journal of pharmacy and pharmacology, 61(7), 883-890 (2009-07-11)
The aim was to establish a preclinical in-vitro system of the nasal mucosa for the simultaneous evaluation of nasal absorption and effects on ciliary activity. Human nasal epithelial cells were grown in collagen-coated transport inserts with transparent polyethylene terephthalate membranes
A Peñalver et al.
Journal of chromatography. A, 839(1-2), 253-260 (1999-05-18)
Three pesticides usually added to paint formulations, Irgarol 1051, dichlofluanid and 4-chloro-3-methylphenol, were determined by solid-phase microextraction (SPME) with 85-micron polyacrylate fibers and gas chromatography-mass spectrometry. The parameters affecting the SPME process (the pH, the addition of salt to the
Kathryn D Baker et al.
Behavioural brain research, 206(1), 143-146 (2009-09-01)
Young chicks were trained on a weakly reinforced variant of a single-trial discrimination avoidance task which typically fails to consolidate the long-term memory stage. The ryanodine receptor (RyR) agonist 4-chloro-m-cresol (500 microM, i.c.) persistently promoted high retention until at least
Niels Ørtenblad et al.
The Journal of physiology, 589(Pt 3), 711-725 (2010-12-08)
Glucose is stored as glycogen in skeletal muscle. The importance of glycogen as a fuel during exercise has been recognized since the 1960s; however, little is known about the precise mechanism that relates skeletal muscle glycogen to muscle fatigue. We
Ramón Jiménez-Moreno et al.
Pflugers Archiv : European journal of physiology, 459(5), 725-735 (2010-01-14)
The endoplasmic/sarcoplasmic reticulum (ER/SR) plays a crucial role in cytoplasmic signalling in a variety of cells. It is particularly relevant to skeletal muscle fibres, where this organelle constitutes the main Ca2+ store for essential functions, such as contraction. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service