Merck
  • A ryanodine receptor agonist promotes the consolidation of long-term memory in young chicks.
A ryanodine receptor agonist promotes the consolidation of long-term memory in young chicks.
Behavioural brain research (2009-09-01)
Kathryn D Baker, Thomas M Edwards, Nikki S Rickard
ABSTRACT
Young chicks were trained on a weakly reinforced variant of a single-trial discrimination avoidance task which typically fails to consolidate the long-term memory stage. The ryanodine receptor (RyR) agonist 4-chloro-m-cresol (500 microM, i.c.) persistently promoted high retention until at least 24 h post-training when administered between the time of training and 20 min post-training. The consolidation of the long-term memory stage by RyR activation implicates intracellular calcium release in triggering long-term memory.
Sigma-Aldrich
4-Chloro-3-methylphenol, ≥98.0% (HPLC)
Sigma-Aldrich
4-Chloro-3-methylphenol, 99%
Supelco
4-Chloro-3-methylphenol, analytical standard