Skip to Content
Merck
All Photos(1)

Documents

24500

Sigma-Aldrich

1-Chlorododecane

≥97.0% (GC)

Synonym(s):

Dodecyl chloride, Lauryl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11Cl
CAS Number:
Molecular Weight:
204.78
Beilstein:
635778
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

form

liquid

transition temp

solidification point -15--10 °C

density

0.867 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCCCCCl

InChI

1S/C12H25Cl/c1-2-3-4-5-6-7-8-9-10-11-12-13/h2-12H2,1H3

InChI key

YAYNEUUHHLGGAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingzhu Liu et al.
Nature materials, 19(12), 1354-1361 (2020-07-29)
Assembling colloidal particles using site-selective directional interactions into predetermined colloidal superlattices with desired properties is broadly sought after, but challenging to achieve. Herein, we exploit regioselective depletion interactions to engineer the directional bonding and assembly of non-spherical colloidal hybrid microparticles.
M A Patrick et al.
Journal of bacteriology, 119(1), 76-81 (1974-07-01)
The effects of hydrocarbons and hydrocarbon derivatives as growth substrates on the polar lipid fractions of an Acinetobacter isolate were studied. Tetradecane, hexadecane, and octadecane resulted in the incorporation of substantial quantities of equivalent-chain-length fatty acids into cellular lipids. Cells
Andrei Filippov et al.
Magnetic resonance in chemistry : MRC, 53(7), 493-497 (2015-04-10)
We used (1) H nuclear magnetic resonance pulsed-field gradient to study the self-diffusion of polyethylene glycol (PEG) and ions in a mixture of PEG and imidazolium bis(mandelato)borate ionic liquids (ILs) at IL concentrations from 0 to 10 wt% and temperatures from
Paniz Sheikholeslami et al.
Experimental eye research, 137, 18-31 (2015-06-06)
The potential of hydrophobically-modified poly(vinyl pyrrolidone) as a shear-responsive, self-associative hydrogel for ophthalmic applications is demonstrated. Hydrophobic modification was achieved via random copolymerization of N-vinylpyrrolidone with N-vinylformamide, the latter of which can be hydrolyzed to expose a desired degree of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service