Skip to Content
Merck
All Photos(3)

Key Documents

185582

Sigma-Aldrich

N,N-Dimethylbenzylamine

≥99%

Synonym(s):

N-Benzyldimethylamine, BDMA, DMBA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N(CH3)2
CAS Number:
Molecular Weight:
135.21
Beilstein:
1099620
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

liquid

refractive index

n20/D 1.501 (lit.)

bp

183-184 °C/765 mmHg (lit.)

mp

−75 °C (lit.)

solubility

water: soluble

density

0.9 g/mL at 25 °C (lit.)

SMILES string

CN(C)Cc1ccccc1

InChI

1S/C9H13N/c1-10(2)8-9-6-4-3-5-7-9/h3-7H,8H2,1-2H3

InChI key

XXBDWLFCJWSEKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylbenzylamine is commonly used as a reagent in organic synthesis and also serves as a catalyst in the synthesis of polyurethane foams and epoxy resins. It reacts with Os3(CO)12 to form triosmium clusters. Anodic oxidation of N,N-dimethylbenzylamine has been studied in methanol-tetra-n-butylammonium fluoroborate and in methanol-potassium hydroxide.

Application

N,N-Dimethylbenzylamine was used in the synthesis of bis[(N,N-dimethylamino)benzyl] selenide. It has been used as catalyst during curing reaction of formulations of diglycidyl ether of bisphenol A and tetrahydrophthalic anhydride.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Indirect ortho functionalization of substituted toluenes through ortho olefination of N, N-dimethylbenzylamines tuned by the acidity of reaction conditions
G Cai, et al.
Journal of the American Chemical Society, 129, 7666-7673 (2007)
Synthesis of waterborne polyurethane acrylate-modified epoxy resin
H Liu, et al.
IOP Conference Series: Materials Science and Engineering, 770, 012062-012062 (2020)
Products and mechanisms in the anodic oxidation of N, N-dimethylbenzylamine in methanol.
Barry JE, et al.
The Journal of Organic Chemistry, 39(18), 2695-2699 (1974)
Mirjana Rodošek et al.
Polymers, 12(3) (2020-03-11)
Versatile product protective coatings that deliver faster drying times and shorter minimum overcoat intervals that enable curing at faster line speeds and though lower energy consumption are often desired by coating manufacturers. Product protective coatings, based on silsesquioxane-modified diglycidyl ether
Wilson Beita-Sandí et al.
Water research, 170, 115323-115323 (2019-12-04)
In this work, we investigated the effect of bromide ion (Br-) on NDMA formation using model precursor compounds, wastewater effluents and surface waters. Previous studies showed that Br- reacts with chloramines and forms bromochloramine, a reactive compound responsible for NDMA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service