Skip to Content
Merck
All Photos(3)

Documents

229628

Sigma-Aldrich

Copper(I) chloride

≥99.995% trace metals basis

Synonym(s):

Copper monochloride, Cuprous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCl
CAS Number:
Molecular Weight:
99.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1.3 mmHg ( 546 °C)

Quality Level

Assay

≥99.995% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: copper

technique(s)

mass spectrometry (MS): suitable

impurities

≤50.0 ppm Trace Rare Earth Analysis

bp

1490 °C (lit.)

mp

430 °C (lit.)

solubility

slightly soluble 0.47 g/L at 20 °C

application(s)

battery manufacturing

SMILES string

Cl[Cu]

InChI

1S/ClH.Cu/h1H;/q;+1/p-1

InChI key

OXBLHERUFWYNTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

The structure of copper(I) chloride is similar to zinc-blende crystal at room temperature; the structure is wurtzite at 407 °C and at higher temperatures it forms copper(I) chloride vapor as per mass spectroscopy.

Application

CuCl may be used as an initiator for hydrostannation of α,α-unsaturated ketones and other similar radical reactions.
Shows unique character as an initiator of radical reactions such as the hydrostannation of α,β-unsaturated ketones.
Shows unique character as an initiator of radical reactions such as the hydrostannation of α,β-unsaturated ketones.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changjun Park et al.
Polymers, 12(2) (2020-02-07)
Here we report the dual light- and thermo-responsive behavior of well-defined rod-coil block copolymers composed of an azobenzene unit, 2-(2-methoxyethoxy)ethyl methacrylate (MEO2MA) and oligo(ethylene glycol) methacrylate (OEGMA). Azobenzene-containing rigid rod blocks prepared by chain growth condensation polymerization of the azobenzene
Madelung O
Semiconductors: Data Handbook null
Yi-Fan Zhao et al.
Journal of colloid and interface science, 448, 380-388 (2015-03-11)
Here we describe the development of versatile antifouling polyethersulfone (PES) filtration membranes modified via surface grafting of zwitterionic polymers from a reactive amphiphilic copolymer additive. Amphiphilic polyethersulfone-block-poly(2-hydroxyethyl methacrylate) (PES-b-PHEMA) was beforehand designed and used as the blending additive of PES
Qidong Wu et al.
ACS omega, 5(36), 23450-23459 (2020-09-22)
Poly(vinylidene fluoride) (PVDF) is a common and inexpensive polymeric material used for membrane fabrication, but the inherent hydrophobicity of this polymer induces severe membranes fouling, which limits its applications and further developments. Herein, we prepared superwettable PVDF membranes by selecting
Yin-Hua Shih et al.
Head & neck, 35(2), 250-256 (2012-03-01)
Lysyl oxidase (LOX) is a copper-dependent enzyme that cross-links collagen and elastin in the extracellular matrix. LOX overexpressed in various tumors. The manner in which LOX affects tumor growth remains controversial. Chemical treatment and gene transfection were used to induce

Articles

Oxidation and reduction reactions are some of the most common transformations encountered in organic synthesis, and are some of the organic chemist’s most powerful tools for creating novel products. Below is a list of the most commonly used oxidizing and reducing agents currently available in our catalog.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

See All

Protocols

We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.

We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service