Skip to Content
Merck
All Photos(1)

Key Documents

451665

Sigma-Aldrich

Copper(II) chloride

anhydrous, powder, ≥99.995% trace metals basis

Synonym(s):

Cupric chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCl2
CAS Number:
Molecular Weight:
134.45
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Assay

≥99.995% trace metals basis

form

powder

impurities

≤50.0 ppm Trace Metal Analysis

mp

620 °C (lit.)

density

3.386 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

Cl[Cu]Cl

InChI

1S/2ClH.Cu/h2*1H;/q;;+2/p-2

InChI key

ORTQZVOHEJQUHG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

CuCl2 was used in studies investigating pleural fibrosis resulting from chronic inhalation of indium phosphide (InP) particles in mice.

Application

Effective catalyst for the tetrahydropyranylation of alcohols, using mild conditions and in high yields.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bhalerao, U.T. et al.
Synthetic Communications, 26, 3081-3081 (1996)
Julia Mazurków et al.
Sensors (Basel, Switzerland), 21(3) (2021-02-04)
The constant progress in novel nanomaterials synthesis has contributed to the rapid development of nonenzymatic glucose sensors. For working electrodes preparation, drop casting proved to be the most convenient and thus most widely applied method. However, appropriate interpretation of obtained
Jing Li et al.
ChemSusChem, 12(19), 4471-4479 (2019-08-02)
A simple one-pot method has been developed to synthesize a palladium/cuprous oxide-copper (Pd/Cu2 O-Cu) material with a well-defined structure, by modification of Cu2 O-Cu with Pd through a galvanic replacement reaction. Compared with the well-known copper/cuprous oxide (Cu/Cu2 O) catalysts
Chen Hou et al.
Nanomaterials (Basel, Switzerland), 10(3) (2020-03-04)
In this work, magnetic CuFe2O4/Ag nanoparticles activated by porous covalent organic frameworks (COF) was fabricated to evaluate the heterogenous reduction of 4-nitrophenol (4-NP). The core-shell CuFe2O4/Ag@COF was successfully prepared by polydopamine reduction of silver ions on CuFe2O4 nanoparticles, followed by
Kenji Funaki et al.
Organic letters, 14(24), 6186-6189 (2012-12-12)
Direct arylation of thiophenes and benzothiophenes with aryltrimethylsilanes was effectively catalyzed by PdCl(2)(MeCN)(2) in the presence of CuCl(2) as an oxidant. The reaction preferentially occurred at the β-position of both thiophenes and benzothiophenes.

Articles

Oxidation and reduction reactions are some of the most common transformations encountered in organic synthesis, and are some of the organic chemist’s most powerful tools for creating novel products. Below is a list of the most commonly used oxidizing and reducing agents currently available in our catalog.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service