Skip to Content
Merck
All Photos(2)

Key Documents

215481

Sigma-Aldrich

Zinc

mossy, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 487 °C)

Assay

≥99%

form

mossy

reaction suitability

core: zinc
reagent type: catalyst

resistivity

5.8 μΩ-cm, 20°C

particle size

1-2 cm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sara Eyal et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 50(5), 798-806 (2009-05-01)
Studies in rodents indicate that the disruption of P-glycoprotein (P-gp) function increases drug distribution into the developing fetus and organs such as the brain. To simultaneously and serially evaluate the effect of P-gp activity and inhibition on the tissue distribution
M E Kuyumcu et al.
European review for medical and pharmacological sciences, 17(11), 1467-1471 (2013-06-19)
Zinc is one of the most important elements for human body. Zinc deficiency can occur in any age, if it is seen in elderly its clinical results can be more harmful due to already diminished functions. Some studies showed zinc
Nalini Pandey et al.
Indian journal of experimental biology, 51(7), 548-555 (2013-08-01)
Black gram plants subjected to varying levels of Zn supply (0.01 to 10 microM Zn) showed optimum growth and dry matter yield in plants receiving 1 microM Zn. The dry matter yield of plants decreased in plants receiving 0.01 and
Casper Groth et al.
Development (Cambridge, England), 140(14), 3018-3027 (2013-06-21)
Developmental patterning requires the precise interplay of numerous intercellular signaling pathways to ensure that cells are properly specified during tissue formation and organogenesis. The spatiotemporal function of the Notch signaling pathway is strongly influenced by the biosynthesis and intracellular trafficking
Bikha Ram Devrajani et al.
JPMA. The Journal of the Pakistan Medical Association, 63(3), 369-373 (2013-08-07)
To determine the serum zinc level in patients with pneumonia. The descriptive cross-sectional study was conducted at the Liaquat University Hospital, Hyderabad, Pakistan, from September 2008 to February 2009. All patients above 12 years of age and of either gender

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service