Skip to Content
Merck
All Photos(2)

Documents

208981

Sigma-Aldrich

Potassium hexachloroiridate(IV)

technical grade

Synonym(s):

Iridium potassium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2IrCl6
CAS Number:
Molecular Weight:
483.13
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Quality Level

form

powder

concentration

≥39.0% Ir (gravimetric)

mp

>385 °C (lit.)

SMILES string

[K+].[K+].Cl[Ir--](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.Ir.2K/h6*1H;;;/q;;;;;;+4;2*+1/p-6

InChI key

WEKZJZDULNNSCD-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nootan Bhattarai et al.
Inorganic chemistry, 51(24), 13303-13311 (2012-11-29)
The aqueous oxidations of glutathione (GSH) by [IrCl(6)](2-), [Fe(bpy)(2)(CN)(2)](+), and [Fe(bpy)(CN)(4)](-) are described. All three reactions are highly susceptible to catalysis by traces of copper ions, but this catalysis can be fully suppressed with suitable chelating agents. The direct oxidation
G M Kolodny et al.
Cancer research, 43(7), 3101-3103 (1983-07-01)
Sodium hexachloroiridate injected 1 day after i.p. injection of 10(6) mouse ovarian tumor cells prevents the appearance of ascitic tumors in mice. Mice given injections of tumor cells all die at 20 to 30 days after tumor injection. Mice treated
V Duarte et al.
Nucleic acids research, 27(2), 496-502 (1998-12-24)
Oxidative damage to DNA bases commonly resultsin the formation of oxidized purines, particularly 7,8-dihydro-8-oxoguanine (8-oxoG) and 7,8-dihydro-8-oxoadenine (8-oxoA), the former being a well-known mutagenic lesion. Since 8-oxoG is readily subject to further oxidation compared with normal bases, the insertion of
Salah Boussaad et al.
Journal of the American Chemical Society, 130(12), 3780-3787 (2008-03-07)
In an effort to develop sensitive nanoscale devices for chemical and biological sensing, we have examined, using liquid gating, the conductance of semiconducting single-walled carbon nanotube-based field-effect transistors (SWCNT-FETs) in the presence of redox mediators. As examples, redox couples K3Fe(CN)6/K4Fe(CN)6
J G Muller et al.
Nucleic acids research, 26(9), 2247-2249 (1998-06-20)
Two gel electrophoretic methods are described for detection of 7, 8-dihydro-8-oxoguanine and 7,8-dihydro-8-oxoadenine based on their further oxidation with one-electron oxidants including IrCl62-and IrBr62-. The products of nucleobase oxidation lead to enhanced piperidine-sensitive cleavage and to highly visible stop points

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service