Skip to Content
Merck
All Photos(3)

Documents

262005

Sigma-Aldrich

Hexaammineruthenium(III) chloride

98%

Synonym(s):

Hexaammineruthenium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Ru(NH3)6]Cl3
CAS Number:
Molecular Weight:
309.61
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

storage temp.

2-8°C

SMILES string

N.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Ru+3]

InChI

1S/3ClH.6H3N.Ru/h3*1H;6*1H3;/q;;;;;;;;;+3/p-3

InChI key

GBDZMMXUOBAJMN-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Hexaammineruthenium III/II (HexRu(III)|HexRu(II)) couple is a commonly used electrochemical redox couple due to its chemical and electrochemical reversibility.

Application

Hexaammineruthenium(III) chloride can be used as a precursor for the preparation of:
  • Ru/Al2O3 catalyst for CO2 hydrogenation.
  • Bimetallic complexes ([Ru(NH3)6][Ag(CN)2]3·2H2O).
  • Chloropentaammineruthenium(III) chloride.
For small scale and high throughput uses, product is also available as ChemBeads (932183)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Measurement of the diffusion coefficients of [Ru (NH3) 6] 3+ and [Ru (NH3) 6] 2+ in aqueous solution using microelectrode double potential step chronoamperometry.
Wang Y, et al,
Journal of Electroanalytical Chemistry, 652(1-2), 13-17 (2011)
Assisted self-association of dicyanoaurate,[Au (CN) 2]-, and dicyanoargentate,[Ag (CN) 2]-, through hydrogen bonding to metal ammonia complexes
Pham D, et al.
Inorganica chimica acta, 358, 4261-4269 (2005)
Juan Yao et al.
The Analyst, 142(21), 4116-4123 (2017-10-11)
An isothermal, enzyme free, ultra-specific and ultra-sensitive protocol for electrochemical detection of miRNAs is proposed based on the toehold-mediated strand displacement reaction (SDR) and non-enzymatic catalytic hairpin reaction (CHA) recycling. The SDR was first triggered only in the presence of
Yi Liu et al.
Science advances, 6(7), eaay5993-eaay5993 (2020-02-29)
Zeolite nanosheets have shown unprecedented opportunities for a wide range of applications, yet developing facile methods for fabrication of uniform zeolite nanosheets remains a great challenge. Here, a facile approach involving anisotropic etching with an aqueous solution of tetrapropylammonium hydroxide
Binuclear complexes of ruthenium ammines.
Creutz C and Taube H
Journal of the American Chemical Society, 95(4), 1086-1094 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service