Skip to Content
Merck
All Photos(1)

Key Documents

202304

Sigma-Aldrich

Poly(propylene glycol)

average Mn ~425

Synonym(s):

PPG, Poly(propylene oxide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H[OCH(CH3)CH2]nOH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
vapor pressure:
<0.01 mmHg ( 20 °C)

vapor density

>1 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

mol wt

average Mn ~425

contains

130-190 ppm proprietary phenolic antioxidant

refractive index

n20/D 1.447

viscosity

80 cSt(25 °C)(lit.)

hydroxyl value

263 mg KOH/g

solubility

water: miscible (completely)

density

1.004 g/mL at 25 °C

SMILES string

CC(O)CO

InChI

1S/C6H14O3/c1-5(8)4-9-6(2)3-7/h5-8H,3-4H2,1-2H3

InChI key

DUFKCOQISQKSAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly(propylene glycol) can be used:
  • As a plasticizer as it does not crystalize, has a low glass transition temperature, and is biocompatible. For example, it can be used in the plasticization of polylactic acid.
  • To prepare the aqueous biphasic system for the recovery of ectoine from Halomonas salina cells.
  • To study the virtual transition from supramolecular polymer(PPG) toconventional polymer(PPO).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

445.0 °F - closed cup

Flash Point(C)

229.44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessalyn Cortese et al.
Journal of the American Chemical Society, 134(8), 3671-3674 (2012-02-11)
We show here that complementary interactions can suppress mesoscopic order and thus lead to a counterintuitive change in material properties. We present results for telechelic supramolecular polymers based on poly(propylene oxide) (PPO), thymine (Thy), and diaminotriazine (DAT). The self-complementary systems
Claudio Poggio et al.
Journal of endodontics, 38(9), 1239-1243 (2012-08-16)
The objective of this study was to evaluate and compare in vitro the decalcifying capability on root canal dentin of 4 different irrigating solutions. Twenty-five freshly extracted maxillary central incisors were selected. The canals were prepared to obtain a total
Zhiyong Zhao et al.
Chemical communications (Cambridge, England), 48(78), 9753-9755 (2012-08-25)
The designed DNA sequences can make DNA-b-PPO undergo in situ transition between diblock and triblock upon pH changes, consequently, induce a morphology-shifting from spherical micelles to nanofibers. This process is reversible and the assembled structures have been characterized by CD
Celeste R Brennecka et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 100(5), 1298-1309 (2012-04-20)
This study examines the in vitro characteristics of a crosslinking polymer system for cerebral aneurysm embolization. The polymeric material is composed of poly(propylene glycol)diacrylate (PPODA) and pentaerythritol tetrakis(3-mercaptopropionate) (QT), formulated with the liquid contrast agents Conray™ or Omnipaque™ 300. The
Chih Hao Chang et al.
Bio-medical materials and engineering, 22(6), 373-382 (2012-11-02)
Segmented polyurethane (SPU) materials based on different soft-segment component (PPG, PTMO and PBA) and various length of soft-segment (molecular weight of PBA: 500, 700 and 1000) were synthesized in this research. The soft-segment components were synthesized from polyether-polyols (PPG and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service