Merck
  • pH-induced morphology-shifting of DNA-b-poly(propylene oxide) assemblies.
pH-induced morphology-shifting of DNA-b-poly(propylene oxide) assemblies.
Chemical communications (Cambridge, England) (2012-08-25)
Zhiyong Zhao, Liying Wang, Yu Liu, Zhongqiang Yang, Yan-Mei He, Zhibo Li, Qing-Hua Fan, Dongsheng Liu
ABSTRACT
The designed DNA sequences can make DNA-b-PPO undergo in situ transition between diblock and triblock upon pH changes, consequently, induce a morphology-shifting from spherical micelles to nanofibers. This process is reversible and the assembled structures have been characterized by CD, TEM and fluorescent experiments.
Sigma-Aldrich
Poly(propylene glycol), average Mn ~2,000
Sigma-Aldrich
Poly(propylene glycol), average Mn ~4,000
Sigma-Aldrich
Poly(propylene glycol), average Mn ~1,000
Sigma-Aldrich
Poly(propylene glycol), average Mn ~425
Sigma-Aldrich
Poly(propylene glycol), average Mn ~2,700
Sigma-Aldrich
Poly(propylene glycol), average Mn ~725
Sigma-Aldrich
Polypropylene glycol, P 2,000
Sigma-Aldrich
Polypropylene glycol, P 400