Skip to Content
Merck
All Photos(4)

Documents

201626

Sigma-Aldrich

Fluoresceinamine, isomer I

Synonym(s):

5-Aminofluorescein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H13NO5
CAS Number:
Molecular Weight:
347.32
Beilstein:
48395
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

mp

223 °C (dec.) (lit.)

solubility

methanol: 5 mg/mL

λmax

496 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Nc1ccc2c(c1)C(=O)OC23c4ccc(O)cc4Oc5cc(O)ccc35

InChI

1S/C20H13NO5/c21-10-1-4-14-13(7-10)19(24)26-20(14)15-5-2-11(22)8-17(15)25-18-9-12(23)3-6-16(18)20/h1-9,22-23H,21H2

InChI key

GZAJOEGTZDUSKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fluoresceinamine, isomer I belongs to the class of derivatized fluoresceins.

Application

Fluoresceinamine, isomer I is suitable for use in a specific and sensitive spectrophotometric method for determining nitrite. It has been used to fluorescently tag nanoparticles through a competitive carboxyl-amine coupling reaction to visualize nanoparticle internalization.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Gribbon et al.
Biophysical journal, 77(4), 2210-2216 (1999-10-08)
Hyaluronan (HA) is a highly hydrated polyanion, which is a network-forming and space-filling component in the extracellular matrix of animal tissues. Confocal fluorescence recovery after photobleaching (confocal-FRAP) was used to investigate intramolecular hydrogen bonding and electrostatic interactions in hyaluronan solutions.
pH sensor based on immobilized fluoresceinamine
Saari L A
Analytical Chemistry, 54, 821-823 (1982)
Mirko Nitschke et al.
Colloids and surfaces. B, Biointerfaces, 90, 41-47 (2011-10-22)
Physico-chemical and topographical cues allow to control the behavior of adherent cells. Towards this goal, commercially available cell culture carriers can be finished with a laterally microstructured biomolecular functionalization. As shown in a previous study [Biomacromolecules 4 (2003) 1072], the
D M Kranz et al.
The Journal of biological chemistry, 257(12), 6987-6995 (1982-06-25)
Binding of fluorescyl ligand by five IgG anti-fluorescyl hybridoma proteins (4-4-20, 6-10-6, 20-4-4, 20-19-=1, 20-20-3) was examined. Relative reduction in fluorescence of bound fluorescein, deuterium oxide (D2O)-induced enhancement of fluorescence, and the effects of pH on binding kinetics were measured
S Miyauchi et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 12(1), 27-34 (1996-01-01)
The protective efficacy of sodium hyaluronate (Na-HA) on the corneal endothelium against the damage induced by sonication was investigated using enucleated rabbit eyes and Na-HA fluorescence labeled with 5-aminofluorescein (FA-HA). The anterior chamber was reformed by injecting a 1% solution

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service