Skip to Content
Merck
All Photos(3)

Key Documents

186279

Sigma-Aldrich

5-Aminoisophthalic acid

94%

Synonym(s):

5-Aminobenzene-1,3-dicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3-1,3-(CO2H)2
CAS Number:
Molecular Weight:
181.15
Beilstein:
2805628
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

94%

reaction suitability

reaction type: solution phase peptide synthesis

mp

>300 °C (lit.)

application(s)

peptide synthesis

SMILES string

Nc1cc(cc(c1)C(O)=O)C(O)=O

InChI

1S/C8H7NO4/c9-6-2-4(7(10)11)1-5(3-6)8(12)13/h1-3H,9H2,(H,10,11)(H,12,13)

InChI key

KBZFDRWPMZESDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Aminoisophthalic acid (AIPA) can be used as a starting material to prepare:
  • Poly(5-aminoisophthalic acid) by oxidative polymerization reaction.
  • Poly(benzimidazole-co-aniline) (PBIANI) by condensation polymerization with 3,3′-diaminobenzidine via poly(5-aminoisophthalic acid) as a key intermediate.

It can also be used as a blended material with polyvinylalcohol to prepare silica-functionalized composite membranes for diffusion dialysis applications. AIPA based metal-organic framework (MOF) can be modified with various functional groups for different applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel silica-functionalized aminoisophthalic acid-based membranes for base recovery via diffusion dialysis
Mondal AN, et al.
Journal of Membrane Science , 507(1-2), 90-98 (2016)
A copper based metal-organic framework: Synthesis, modification and VOCs adsorption
Xu Wei-Qin, et al.
Inorganic Chemistry Communications, 92(1-2), 1-4 (2018)
En Tang et al.
Inorganic chemistry, 45(16), 6276-6281 (2006-08-02)
Two stable supramolecular microporous cobalt(II) polymers, namely [Co(HAIP)2]n.3nH2O (1) and [Co(AIP)(H2O)]n (2), AIP = 5-aminoisophthalate, were hydrothermally synthesized and characterized by single-crystal X-ray diffraction, IR spectra, thermogravimetric analyses, and variable-temperature magnetic susceptibility measurements. The two complexes are constructed from the
A new self-cross-linked, net-structured, proton conducting polymer membrane for high temperature proton exchange membrane fuel cells
Bhadra S, et al.
Journal of Membrane Science , 349(1-2), 304-311 (2010)
D Lecouturier et al.
Applied microbiology and biotechnology, 62(5-6), 550-556 (2003-07-08)
5-Amino-2,4,6-triiodoisophthalic acid (ATIA), both a precursor and a degradative intermediate of triiodinated contrast media, was anaerobically converted by sludge from a wastewater treatment plant. ATIA conversion took place only when an electron donor such as ethanol was added. A stable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service