Skip to Content
Merck
All Photos(4)

Documents

117374

Sigma-Aldrich

4′-Methoxyacetophenone

99%

Synonym(s):

4-Acetylanisole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4COCH3
CAS Number:
Molecular Weight:
150.17
Beilstein:
742313
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39023808
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

bp

152-154 °C/26 mmHg (lit.)

mp

36-38 °C (lit.)

solubility

H2O: soluble 2.474 g/L at 20 °C

SMILES string

COc1ccc(cc1)C(C)=O

InChI

1S/C9H10O2/c1-7(10)8-3-5-9(11-2)6-4-8/h3-6H,1-2H3

InChI key

NTPLXRHDUXRPNE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

4′-Methoxyacetophenone undergoes biocatalytic enantioselective reduction using immobilized Rhodotorula sp. AS2.2241 cells to yield (S)-1-(4-methoxyphenyl) ethanol in a hydrophilic ionic liquid-containing co-solvent system.

Application

4′-Methoxyacetophenone was used to study ruthenium catalyzed step growth copolymerization of 4′-methoxyacetophenone with α,ω-dienes to yield copolymers. It was used to determine the activity of various ketones approved as food additives against Clostridium Botulinum spores and cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

280.4 °F - open cup

Flash Point(C)

138 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruthenium catalyzed regioselective step-growth copolymerization of 4'-methoxyacetophenone, or 4'-phenoxyacetophenone and a, ?-dienes.
Guo H and Weber WP.
Polym. Bull., 35(3), 259-264 (1995)
Antibotulinal properties of selected aromatic and aliphatic ketones.
Bowles BL and Miller AJ.
Journal of Food Protection, 56(9), 795-800 (1993)
[Toxicological characteristics of acetylanisole].
M I Makaruk et al.
Gigiena i sanitariia, (4)(4), 86-87 (1985-04-01)
Wen-Yong Lou et al.
Journal of biotechnology, 143(3), 190-197 (2009-07-21)
The biocatalytic enantioselective reduction of 4'-methoxyacetophenone to (S)-1-(4-methoxyphenyl)ethanol was successfully conducted in a hydrophilic IL-containing co-solvent system using immobilized Rhodotorula sp. AS2.2241 cells. Of all the tested ILs, the best results were observed with the novel IL 1-(2'-hydroxy)ethyl-3-methylimidazolium nitrate (C(2)OHMIM.NO(3))
Sanja Ćavar et al.
Pharmaceutical biology, 48(2), 170-176 (2010-07-22)
Stachys menthifolia Vis. (Lamiaceae) is an endemic species from the Balkan Peninsula spread throughout Albania, Greece, Montenegro, and Croatia. This article presents the first investigation of the essential oil composition of this species from Croatia. Aerial parts of the plant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service