Skip to Content
Merck
All Photos(1)

Key Documents

165336

Sigma-Aldrich

Methylboronic acid

97%

Synonym(s):

Methaneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3B(OH)2
CAS Number:
Molecular Weight:
59.86
Beilstein:
1731087
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

91-94 °C (lit.)

SMILES string

CB(O)O

InChI

1S/CH5BO2/c1-2(3)4/h3-4H,1H3

InChI key

KTMKRRPZPWUYKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methylboronic acid can be used as a reagent:
  • In the palladium-catalyzed Stille and Suzuki-Miyaura cross-couplings.
  • In the microwave-heated heterogeneous palladium (Pd)-catalytized reactions.
  • In ruthenium (Ru)-catalyzed silylation reactions
  • To prepare bis(aminotropone) titanium (Ti) catalysts for ethylene polymerizations.
  • In the enantioselective asymmetric bromoaminocyclization and bromoaminocyclization using amino-thiocarbamate catalysts.
  • To prepare common building blocks for pharmaceuticals and agrochemicals.
  • To prepare chrysin analogs by Suzuki-Miyaura coupling reactions.
  • To prepare casein kinase I inhibitors.
  • In the divergent C-H functionalizations directed by sulfonamide pharmacophores in drug discovery.
  • In the synthesis of unsymmetrical monosulfides from disulfides via copper-catalyzed coupling with boronic acids.
  • In a palladium-catalyzed coupling with enol tosylates.
  • For derivatizing many carbohydrates and biologically active compounds for GLC analysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hui-Xiong Dai et al.
Journal of the American Chemical Society, 133(18), 7222-7228 (2011-04-15)
Modern drug discovery is contingent on identifying lead compounds and rapidly synthesizing analogues. The use of a common pharmacophore to direct multiple and divergent C-H functionalizations of lead compounds is a particularly attractive approach. Herein, we demonstrate the viability of
Ruthenium-catalyzed C-H silylation of methylboronic acid using a removable a-directing modifier on the boron atom
Ihara, H.; Ueda, A.; Suginome, M.
Chemistry Letters (Jpn), 40, 916-918 (2011)
Chemical Development of the Casein Kinase I - Epsilon Inhibitor: 3-(3-Fluorophenyl)sulfanyl-1H-pyrrolo[3,2-b]pyridine-2-carboxylic Acid Amide
Huang, B-G.; et al.
Organic Process Research & Development, 15, 1040-1045 (2011)
Selective heating of Pd-modified ordered mesoporous carbon CMK-3 by microwave irradiation
Inagaki, S.; et al.
Bulletin of the Chemical Society of Japan, 84, 1136-1143 (2011)
Ling Zhou et al.
Journal of the American Chemical Society, 133(24), 9164-9167 (2011-05-05)
A facile and efficient enantioselective bromoaminocyclization of unsaturated sulfonamides has been developed using an amino-thiocarbamate catalyst. A range of enantioenriched pyrrolidines were prepared with up to 99% yield and 99% ee. The corresponding lactams could be obtained through oxidation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service