Skip to Content
Merck
All Photos(4)

Key Documents

160601

Sigma-Aldrich

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p′-disulfonic acid monosodium salt hydrate

97%

Synonym(s):

FerroZine Iron Reagent, PDT disulfonate monosodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H13N4NaO6S2 · xH2O
CAS Number:
Molecular Weight:
492.46 (anhydrous basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
solid

grade

for analytical purposes

Assay

97%

form

solid

functional group

sulfonic acid

SMILES string

O.[Na+].OS(=O)(=O)c1ccc(cc1)-c2nnc(nc2-c3ccc(cc3)S([O-])(=O)=O)-c4ccccn4

InChI

1S/C20H14N4O6S2.Na.H2O/c25-31(26,27)15-8-4-13(5-9-15)18-19(14-6-10-16(11-7-14)32(28,29)30)23-24-20(22-18)17-3-1-2-12-21-17;;/h1-12H,(H,25,26,27)(H,28,29,30);;1H2/q;+1;/p-1

InChI key

MBOKSYFCYXIJRZ-UHFFFAOYSA-M

Application

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p′-disulfonic acid monosodium salt hydrate has been used as a colorimetric indicator for ferroxidase assay.
Reagent for the determination of Fe and Co, and Ag, Cu, and Ni.

Legal Information

FerroZine is a trademark of Hach Company

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of iron-related proteins at the neurovascular unit supports reduction and reoxidation of iron for transport through the blood-brain barrier.
Burkhart A, et al.
Molecular Neurobiology, 53(10), 7237-7253 null
G Al-Jabari et al.
Talanta, 34(5), 479-482 (1987-05-01)
Silver(I), copper(II) and nickel(II) can be reduced to the metallic state by formaldehyde at pH 11, chromium(II) in 2.5M sulphuric acid, and borohydride at pH 5.5-6.0, respectively. Reoxidation of these metals with iron(III) in the presence of Ferrozine enables their
Chandrika N Deshpande et al.
PloS one, 12(9), e0184366-e0184366 (2017-09-08)
Hephaestin is a large membrane-anchored multicopper ferroxidase involved in mammalian iron metabolism. Newly absorbed dietary iron is exported across the enterocyte basolateral membrane by the ferrous iron transporter ferroportin, but hephaestin increases the efficiency of this process by oxidizing the
Pao-Yuan Hsiao et al.
Scientific reports, 7(1), 9175-9175 (2017-08-25)
Plant defensins (PDFs) are cysteine-rich peptides that have a range of biological functions, including defence against fungal pathogens. However, little is known about their role in defence against bacteria. In this study, we showed that the protein encoded by ARABIDOPSIS
M V Dawson et al.
Talanta, 37(12), 1189-1191 (1990-12-01)
Procedures are described whereby iron (1-50 microg) and cobalt (1-25 microg) are determined spectrophotometrically, iron as iron(II) with the disodium salt of 3-(2-pyridyl)-5,6-bis(4-phenylsulfonic acid)-1,2,4-triazine (Ferrozine) and cobalt as the cobalt(III) dithizonate complex. The reduction to iron(II) prevents interference of iron(III)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service