Skip to Content
Merck
All Photos(1)

Key Documents

15500

Sigma-Aldrich

Boc-Ser-OH

≥99.0% (T), for peptide synthesis

Synonym(s):

N-(tert-Butoxycarbonyl)-L-serine, Boc-L-serine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
205.21
Beilstein:
2212252
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

product name

Boc-Ser-OH, ≥99.0% (T)

Assay

≥99.0% (T)

optical activity

[α]20/D −3.5±0.5°, c = 2% in acetic acid

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

91 °C (dec.) (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N[C@@H](CO)C(O)=O

InChI

1S/C8H15NO5/c1-8(2,3)14-7(13)9-5(4-10)6(11)12/h5,10H,4H2,1-3H3,(H,9,13)(H,11,12)/t5-/m0/s1

InChI key

FHOAKXBXYSJBGX-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Boc-Ser-OH is an amino acid derivative.

Application

Boc-Ser-OH may be used in the synthesis of the following:
  • 2-(N-Fmoc)-3-(N-Boc-N-methoxy)-diaminopropanoic acid (Fmoc: 9-fluorenylmethoxycarbonyl; Boc: t-butyloxycarbonyl)
  • Boc-Ser-Leu-OMe
  • cyclic peptide synthesis
  • benzylsulfonyl-D-Ser-Ser-4-amidinobenzylamide
Building block in peptide synthesis. Starting material for the synthesis of various α-amino acids via the β-lactone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Synthesis of cyclic peptides via O-N-acyl migration.
Lecaillon J, et al.
Tetrahedron Letters, 49(31), 4674-4676 (2008)
O García-Suárez et al.
Immunology, 94(2), 235-241 (1998-09-19)
Increasing evidence suggests that some members of the neurotrophic factor family of neurotrophins could be implicated in the regulation of immune responses. Neurotrophins, as well as their tyrosine kinase signal-transducing receptors (the so-called Trk neurotrophin receptors), have been detected in
Gema Villa-Fombuena et al.
Development (Cambridge, England), 148(18) (2021-08-10)
Drosophila female germline stem cells (GSCs) are found inside the cellular niche at the tip of the ovary. They undergo asymmetric divisions to renew the stem cell lineage and to produce sibling cystoblasts that will in turn enter differentiation. GSCs
Michael R Carrasco et al.
Biopolymers, 84(4), 414-420 (2006-03-02)
Amino acids with N-alkylaminooxy side chains have proven effective for the rapid synthesis of neoglycopeptides. Chemoselective reaction of reducing sugars with peptides containing these amino acids provides glycoconjugates that are structurally similar to their natural counterparts. 2-(N-Fmoc)-3-(N-Boc-N-methoxy)-diaminopropanoic acid (Fmoc: 9-fluorenylmethoxycarbonyl;
M Komai-Koma et al.
Immunology, 91(4), 579-585 (1997-08-01)
The chemoattractant effect of soluble protein antigens for B cells from immunized mice was examined. Mice were immunized either via the footpad with ovalbumin (OVA) in complete Freund's adjuvant (CFA) or with CFA alone; or intraperitoneally with OVA incorporated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service