
Boc-Ser-OH

Synonym(s):
N-(tert-Butoxycarbonyl)-L-serine, Boc-L-serine, Boc-Ser-OH
Linear Formula:
HOCH2CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
205.21
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing