All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C3H7NS
CAS Number:
Molecular Weight:
89.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
form
liquid
refractive index
n20/D 1.5508 (lit.)
bp
72-75 °C/25 mmHg (lit.)
density
1.131 g/mL at 25 °C (lit.)
functional group
thioether
SMILES string
C1CSCN1
InChI
1S/C3H7NS/c1-2-5-3-4-1/h4H,1-3H2
InChI key
OGYGFUAIIOPWQD-UHFFFAOYSA-N
Application
Thiazolidine was used in the synthesis of homogeneous penicillamine disulphide cross-linked polypeptides.
Signal Word
Warning
Hazard Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
132.8 °F - closed cup
Flash Point(C)
56 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
David Pertuit et al.
Natural product communications, 10(6), 1005-1007 (2015-07-23)
A new aromatic compound 3,4,5-trimethoxyphenyl-1-O-(4-sulfo)-β-D-glucopyranoside (1), in addition to two triterpenoid saponins (chebuloside II, arjunoglucoside II), two triterpenes (arjunolic acid and 3-betulinic acid) and sitosterol-3-O-β-D-glucopyranoside have been isolated from the barks of Terminalia catappa. Their structures have been established on
Sinem Aslan Erdem et al.
Phytochemistry, 110, 160-165 (2014-12-20)
Four new oleanane-type saponins 3-O-α-L-rhamnopyranosyl-(1 → 4)-β-D-glucuronopyranosyl-22-O-β,β-dimethylacryloylA1-barrigenol (1), 3-O-α-L-rhamnopyranosyl-(1 → 4)-β-D-glucuronopyranosyl-22-O-angeloylA1-barrigenol (2), 3-O-β-D-glucopyranosyl-(1 → 2)-[β-D-glucopyranosyl-(1 → 6)]-β-D-glucopyranosyl-21,22,28-O-triacetyl-(3β,21β,22α)-olean-12-en-16-one (3), and 3-O-β-D-glucopyranosyl-(1 → 2)-glucopyranosyl-22-O-β-D-glucopyranosylsteganogenin (4), along with the known 3-O-β-D-galactopyranosyl-(1 → 2)-[α-L-arabinopyranosyl-(1 → 3)]-β-D-glucuronopyranosyl-22-O-angeloylA1-barrigenol and 3-O-α-L-rhamnopyranosyl-(1 → 4)-β-D-glucuronopyranosyloleanolic acid, were isolated from
Mark D Ericson et al.
Tetrahedron letters, 54(26), doi:10-doi:10 (2013-12-19)
The syntheses of homogeneous penicillamine disulfide cross-linked polypeptides are reported. Dodecapeptides containing N-terminal, C-terminal, or N- and C-terminal Pen were serially ligated into 36 amino acid polypeptides linked through Cys-Pen, Pen-Cys or Pen-Pen disulfide bonds. Critical to the syntheses was
Takahiro Sawada et al.
Journal of atherosclerosis and thrombosis, 27(7), 644-656 (2019-10-22)
So far, the mechanisms behind the cardiovascular benefits of sodium/glucose cotransporter 2 (SGLT2) inhibitors have not been fully clarified. In order to evaluate the effects of SGLT2 inhibitors on systemic hemodynamics, glucose metabolism, lipid profile, and endothelial function, 50 diabetic
Nampoina Andriamisaina et al.
Phytochemistry, 160, 78-84 (2019-02-12)
The phytochemical study of Ornithogalum dubium Houtt. (Asparagaceae) led to the isolation of five undescribed steroidal glycosides together with two known ones. Their structures were established by using NMR analysis and mass spectrometry as (25R)-3β-hydroxyspirost-5-en-1β-yl O-α-L-arabinopyranosyl-(1 → 2)-α-L-rhamnopyranoside, (25S)-3β-hydroxyspirost-5-en-1β-yl O-β-D-glucopyranosyl-(1 → 6)-β-D-glucopyranoside, (22S)-16β-[(α-L-rhamnopyranosyl)oxy]-22-hydroxycholest-5-en-3β-yl O-β-D-glucopyranosyl-(1 → 4)-β-D-glucopyranoside
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service