Skip to Content
Merck
All Photos(2)

Key Documents

R7017

Sigma-Aldrich

Resazurin sodium salt

powder, BioReagent

Synonym(s):

7-Hydroxy-3H-phenoxazin-3-one-10-oxide sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6NNaO4
CAS Number:
Molecular Weight:
251.17
Beilstein:
3794971
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

product line

BioReagent

form

powder

composition

Dye content, ~80%

pH

3.8-6.5, orange to violet

solubility

H2O: soluble

λmax

600 nm

SMILES string

[Na+].[O-]c1ccc2c(OC3=CC(=O)C=CC3=[N+]2[O-])c1

InChI

1S/C12H7NO4.Na/c14-7-1-3-9-11(5-7)17-12-6-8(15)2-4-10(12)13(9)16;/h1-6,14H;/q;+1/p-1

InChI key

IVGPGQSSDLDOLH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

For the measurement of metabolic activity and proliferation of living cells. The bioreduction of the dye reduces the amount of the oxidized form (blue) and concomitantly increases the fluorescent intermediate (red).
Resazurin sodium salt has been used in viability assay:
  • to measure cell viability of A. fumigatus
  • to assess cell proliferation
  • to determine A. fumigatus viability in high-throughput screenings of microbial natural extracts

Biochem/physiol Actions

Resazurin is a blue, non-fluorescent dye originally used to test milk/biological fluids for bacterial and yeast contamination. It is also useful in sperm viability and semen quality test. Resazurin is applicable in cytotoxicity determination.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sugandha Bhatia et al.
Journal of clinical medicine, 8(6) (2019-06-27)
Dynamic interconversions between transitional epithelial and mesenchymal states underpin the epithelial mesenchymal plasticity (EMP) seen in some carcinoma cell systems. We have delineated epithelial and mesenchymal subpopulations existing within the PMC42-LA breast cancer cell line by their EpCAM expression. These
Hitting the caspofungin salvage pathway of human-pathogenic fungi with the novel lasso peptide humidimycin (MDN-0010)
Valiante V, et al.
Antimicrobial agents and chemotherapy, 59(9), 5145-5153 (2015)
A new approach to drug discovery: high-throughput screening of microbial natural extracts against Aspergillus fumigatus using resazurin
Monteiro MC, et al.
Journal of Biomolecular Screening, 17(4), 542-549 (2012)
Human umbilical cord tissue-derived mesenchymal stromal cells attenuate remodeling after myocardial infarction by proangiogenic, antiapoptotic, and endogenous cell-activation mechanisms
Nascimento DS, et al.
Stem Cell Research & Therapy, 5(1), 5-5 (2012)
Investigation of the Alamar Blue (resazurin) fluorescent dye for the assessment of mammalian cell cytotoxicity
O Brien J, et al.
European Journal of Biochemistry, 267(17), 5421-5426 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service