Skip to Content
Merck
All Photos(1)

Key Documents

SMB00378

Sigma-Aldrich

Diallyl disulfide

≥98% (HPLC)

Synonym(s):

Allyl disulfide, Diallyl disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2SSCH2CH=CH2
CAS Number:
Molecular Weight:
146.27
Beilstein:
1699241
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

biological source

Allium sativum L.

Quality Level

vapor density

>5 (vs air)

vapor pressure

1 mmHg ( 20 °C)

Assay

≥98% (HPLC)

form

oil

color

colorless to yellow

refractive index

n20/D 1.541 (lit.)

bp

180-195 °C (lit.)

density

1.008 g/mL at 25 °C (lit.)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

C=CCSSCC=C

InChI

1S/C6H10S2/c1-3-5-7-8-6-4-2/h3-4H,1-2,5-6H2

InChI key

PFRGXCVKLLPLIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diallyl disulfide (DADS) is an oil-soluble volatile organosulfur compound that can be naturally found in Allium sativum (garlic). It has a simple chemical structure with two sulfur atoms attached with two allyl groups. This constitute of garlic contributes to its pungent aroma.

Application

Diallyl disulfide has been used:
  • to study its effects in apoptosis induction and cell viability on different glioma cell lines
  • as a treatment and positive control to assess the anti-inflammatory regulation of nuclear factor erythroid-2 related factor-2 (Nrf2) in breast cancer cells
  • as a potential substrate compound to detect hydrogen sulfide (H2S) production via lead acetate/lead sulfide method

Biochem/physiol Actions

Diallyl disulfide is a potent anticancer agent. It can reduce carcinogen-induced cancers and shows anti-proliferative effects in both in vivo and in vitro models. DADS enhances the anti-obesity and anti-inflammatory effects of green tea in high-fat/high-sucrose (HF/HS) diet-fed mice. It also displays antioxidant, anti-microbial, cardiovascular protective, and neuroprotective activities. DADS elicits anti-mitotic and anti-neoplastic effects and arrests mitosis in colon neoplastic lesions in vivo and colon tumor cell growth in vitro experiments.
Organosulfur compound that is naturally found in garlic. Contributes to the pungent odor of crushed garlic. Anticancer agent and may provide protection against cardiovascular disease.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xiuxiu Song et al.
Evidence-based complementary and alternative medicine : eCAM, 2021, 5103626-5103626 (2021-11-09)
Garlic is widely accepted as a functional food and an excellent source of pharmacologically active ingredients. Diallyl disulfide (DADS), a major bioactive component of garlic, has several beneficial biological functions, including anti-inflammatory, antioxidant, antimicrobial, cardiovascular protective, neuroprotective, and anticancer activities.
Jaehoon Bae et al.
The Journal of nutritional biochemistry, 64, 152-161 (2018-12-06)
Obesity is a major problem in developed countries and a burden on social health care systems. Several epidemiological studies showed the protective effects of green tea against obesity-related diseases. Cyclic guanosine monophosphate (cGMP) acts as a mediator for the physiological
Effects of drying time on the aroma of garlic (Allium sativum L.) seasoning powder
Li Kaixuan, et al.,
Flavour and Fragrance Journal, 36(1), 99-110 (2021)
Effects of the garlic compound diallyl disulfide on the metabolism, adherence and cell cycle of HT-29 colon carcinoma cells: evidence of sensitive and resistant sub-populations.
Robert V, et al.
Carcinogenesis, 22(8), 1155-1161 (2001)
Lan Yi et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 57, 362-370 (2013-04-16)
Considerable evidence in recent years suggests that garlic has anti-proliferative effects against various types of cancer. Garlic contains water-soluble and oil-soluble sulfur compounds. Oil-soluble compounds such as diallyl sulfide (DAS), diallyl disulfide (DADS), diallyl trisulfide (DATS) and ajoene are more

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service