Skip to Content
Merck
All Photos(1)

Documents

14508

Sigma-Aldrich

Poly(ethylene glycol) bis(amine)

Mw 10,000, carboxyl reactive, amine

Synonym(s):

Polyethylene glycol, O,O′-Bis(2-aminoethyl)polyethylene glycol, Diaminopolyethylene glycol, PEG-diamine, Polyoxyethylene bis(amine)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2CH2O)nCH2CH2NH2
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

product name

Poly(ethylene glycol) bis(amine), Mw 10,000

mol wt

Mw 10,000

Quality Level

reaction suitability

reagent type: cross-linking reagent
reactivity: carboxyl reactive

Ω-end

amine

α-end

amine

polymer architecture

shape: linear
functionality: homobifunctional

InChI

1S/C6H16N2O2/c7-1-3-9-5-6-10-4-2-8/h1-8H2

InChI key

IWBOPFCKHIJFMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Y Lee et al.
Cancers, 12(9) (2020-09-02)
Acinic cell carcinoma (AcCC) is a morphologically distinctive salivary gland malignancy often associated with chromosome rearrangements leading to overexpression of the NR4A3 transcription factor. However, little is known about how NR4A3 contributes to AcCC biology. Detailed RNA-sequencing of 21 archived
Katia Cortese et al.
Membranes, 10(5) (2020-05-08)
Colorectal cancer (CRC) is one of the main causes of cancer-related death in developed countries. Targeted therapies and conventional chemotherapeutics have been developed to help treat this type of aggressive cancer. Among these, the monoclonal antibodies cetuximab (Cxm) and panitumumab
Christine Romano et al.
Studies in conservation = Etudes de conservation, 65 (2020-02-28)
Zinc (carboxylate) soaps, formed by reactions between zinc oxide (ZnO) and fatty acids in a drying oil, are known to cause deterioration in the paint layers of modern and contemporary oil paintings. This study investigates zinc carboxylates that developed in
Dean F Martin et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 54(10), 1019-1022 (2019-05-11)
Reliable information indicated that a half-million prematurely born babies are routinely given antibiotics without evidence of infection. This study is based on a concern that such antibiotics might go into hospital waste systems and lead to antibiotic-resistant bacteria. We tested
Yufei Zheng et al.
Nutrients, 12(4) (2020-04-03)
The increasing incidence of obesity poses a great threat to public health worldwide. Recent reports also indicate the relevance of obesity in metabolic diseases. Chinese propolis (CP), as a well-studied natural nutraceutical, has shown a beneficial effect on alleviating diabetes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service