Skip to Content
Merck
All Photos(1)

Key Documents

136115

Sigma-Aldrich

Thiocholesterol

Synonym(s):

5-Cholestene-3β-thiol, Cholesteryl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46S
CAS Number:
Molecular Weight:
402.72
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

form

powder

optical activity

[α]20/D −23°, c = 1 in chloroform

mp

97-99 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](S)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C27H46S/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

QGVQZRDQPDLHHV-DPAQBDIFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thiocholesterol is a lipid similar to cholesterol except that the hydroxyl group is replaced with thiol group.

Application

Thiocholesterol is used in biological cross-linking and membrane studies.
It can also be used as a chain transfer agent (CTA) for the radical polymerization of HPMA monolactate and HPMA dilactate monomers to form copolymers. [HPMA=N-(2-hydroxypropyl)methacrylamide]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Tanaka et al.
Lipids, 30(4), 321-325 (1995-04-01)
The effect of thiocholesterol (SH-Chol) on the copper-induced in vitro oxidation of low-density lipoprotein (LDL; 1.019 < d < 1.063) was investigated. Among the antioxidants tested, including cysteine, glutathione, 2-mercaptoethanol, dithiothreitol, probucol, thiopalmitic acid, and SH-Chol, SH-Chol was the most
Kun Cheng et al.
The Journal of pharmacology and experimental therapeutics, 317(2), 797-805 (2006-02-03)
A triplex-forming oligonucleotide (TFO) specific for type alpha1(I) collagen promoter is a promising candidate for treating liver fibrosis. Earlier, we determined the pharmacokinetics and biodistribution of TFO after systemic administration into normal and fibrotic rats. In this study, we conjugated
Zhaohua Huang et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 11(3), 409-417 (2005-02-25)
A series of thiocholesterol-based cationic lipids (TCL) has been designed and synthesized by the attachment of thiocholesterol to a cationic amine via a disulfide bond. TCL can be incorporated into liposomes and used to package DNA into a lipoplex, thereby
G Zhou et al.
Biochimica et biophysica acta, 1258(2), 101-106 (1995-09-14)
Lecithin:cholesterol acyltransferase (LCAT) is a plasma enzyme which catalyses cholesteryl ester formation from lecithin and cholesterol present in the surface of plasma lipoproteins. Sterol fatty acid acceptors have previously been shown to require the presence of a trans conformation of
Avinash Bajaj et al.
Journal of medicinal chemistry, 51(8), 2533-2540 (2008-04-02)
A structure-activity investigation was undertaken to see the effect of the nature of the spacer on the gene transfection efficacies of thiocholesterol-derived cationic gemini lipids possessing disulfide linkage between the cationic headgroup and the thiocholesterol moiety. Three gemini cationic lipids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service