Skip to Content
Merck
All Photos(3)

Documents

H52406

Sigma-Aldrich

3-(4-Hydroxyphenyl)propionic acid

98%

Synonym(s):

3-(p-Hydroxyphenyl)propionic acid, Phloretic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2CH2CO2H
CAS Number:
Molecular Weight:
166.17
Beilstein:
2209841
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

129-131 °C (lit.)

fluorescence

λex 320 nm; λem 404 nm (buffer, pH 8.6)

SMILES string

OC(=O)CCc1ccc(O)cc1

InChI

1S/C9H10O3/c10-8-4-1-7(2-5-8)3-6-9(11)12/h1-2,4-5,10H,3,6H2,(H,11,12)

InChI key

NMHMNPHRMNGLLB-UHFFFAOYSA-N

Application

3-(4-Hydroxyphenyl)propionic acid can be used:
  • To derive hydrogel based on chitosan, gelatin or poly(ethylene glycol).
  • To synthesize novel symmetric and asymmetric bolaamphiphiles.
  • As an acyl donor for the enzymatic synthesis of sugar alcohol esters.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Biocatalytic acylation of sugar alcohols by 3-(4-hydroxyphenyl) propionic acid.
Croitoru R, et al.
Process Biochemistry (Oxford, United Kingdom), 47(12), 1894-1902 (2012)
Efficient syntheses of bolaform surfactants from L-rhamnose and/or 3-(4-hydroxyphenyl) propionic acid.
Akong F O and Bouquillon S
Green Chemistry, 17(6), 3290-3300 (2015)
Seon-Min Jeon et al.
Translational research : the journal of laboratory and clinical medicine, 149(1), 15-21 (2007-01-02)
This study was conducted to compare the hypocholesterolemic and antioxidant effects of naringenin (0.02%), and its metabolites, rho-hyproxyphenylpropionic acid (PHPP, 0.012%) and rho-hydroxybenzoic acid (PHB, 0.012%), in 1% cholesterol diet-fed rats. All supplements significantly lowered the plasma total-cholesterol (total-C), triglyceride
Eliana Capecchi et al.
Nanomaterials (Basel, Switzerland), 8(6) (2018-06-20)
Sustainable catalysts for the oxidation of phenol derivatives under environmentally friendly conditions were prepared by the functionalization of lignin nanoparticles with tyrosinase. Lignin, the most abundant polyphenol in nature, is the main byproduct in the pulp and paper manufacturing industry
Enhanced tissue adhesiveness of injectable gelatin hydrogels through dual catalytic activity of horseradish peroxidase.
Hoang T T T, et al.
Biopolymers, 109(1), e23077-e23077 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service