Skip to Content
Merck
All Photos(1)

Key Documents

135577

Sigma-Aldrich

2-Chlorobenzoic acid

98%

Synonym(s):

2-CBA, o-Chlorobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CO2H
CAS Number:
Molecular Weight:
156.57
Beilstein:
907340
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

138-140 °C (lit.)

solubility

cold water: soluble 900 part
alcohol: freely soluble
diethyl ether: freely soluble
water: soluble (hot)

functional group

carboxylic acid
chloro

SMILES string

OC(=O)c1ccccc1Cl

InChI

1S/C7H5ClO2/c8-6-4-2-1-3-5(6)7(9)10/h1-4H,(H,9,10)

InChI key

IKCLCGXPQILATA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Chlorobenzoic acid degradation by pure cultures of Burkholderia cepacia strain with and without the bacterial hemoglobin gene was studied in parallel membrane bioreactors. Intramolecular hydrogen atom tunneling in 2-chlorobenzoic acid has been studied by low-temperature matrix-isolation infrared spectroscopy.

Application

2-Chlorobenzoic acid was used to study the degradation of 2-bromobenzoic acid by Pseudomonas aeruginosa.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

343.4 °F

Flash Point(C)

173 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

F K Higson et al.
Applied and environmental microbiology, 56(6), 1615-1619 (1990-06-01)
A strain of Pseudomonas aeruginosa producing 2-bromobenzoic acid, designated 2-BBZA, was isolated by enrichment culture from municipal sewage. It degraded all four 2-halobenzoates as well as certain 3-halo- and dihalobenzoates, though none of the 4-halobenzoates supported growth of this organism.
A C Layton et al.
Journal of industrial microbiology, 13(6), 392-401 (1994-11-01)
The microbial populations in PCB-contaminated electric power substation capacitor bank soil (TVA soil) and from another PCB-contaminated site (New England soil) were compared to determine their potential to degrade PCB. Known biphenyl operon genes were used as gene probes in
Gang-hua Lang et al.
Applied microbiology and biotechnology, 83(6), 1085-1094 (2009-03-26)
TfdT is a LysR-type transcriptional regulator that activates the transcription of the chlorocatechol degradative gene operon tfdCDEF of the chlorobenzoate-degrading bacterium Burkholderia sp. NK8. To identify the amino acids involved in the effector recognition by TfdT, a polymerase-chain-reaction-based random mutagenesis
Grazia Baggi et al.
Biodegradation, 16(3), 275-282 (2005-05-04)
A study was made of the metabolic and co-metabolic intermediates of 2- and 3-chlorobenzoate, 2,3- and 3,5-dichlorobenzoate to elucidate the mechanism(s) involved in the negative effects observed on the growth of a chlorobenzoate-degrading microbial consortium in the presence of mixed
Satoshi Nishino et al.
The journal of physical chemistry. A, 111(30), 7041-7047 (2007-07-03)
Intramolecular hydrogen atom tunneling in 2-chlorobenzoic acid has been investigated by low-temperature matrix-isolation infrared spectroscopy with the aid of density functional theory calculation. Infrared spectra of two relatively stable syn isomers, SC and ST, were observed in argon and xenon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service