Skip to Content
Merck
All Photos(1)

Key Documents

13151

Sigma-Aldrich

6-Benzylaminopurine

ReagentPlus®, ≥99.0% (HPLC)

Synonym(s):

6-BAP, BA, N6-Benzyladenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N5
CAS Number:
Molecular Weight:
225.25
Beilstein:
19406
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

≥99.0% (HPLC)

form

solid

ign. residue

≤0.05%

mp

230-233 °C

functional group

amine
phenyl

SMILES string

C(Nc1ncnc2nc[nH]c12)c3ccccc3

InChI

1S/C12H11N5/c1-2-4-9(5-3-1)6-13-11-10-12(15-7-14-10)17-8-16-11/h1-5,7-8H,6H2,(H2,13,14,15,16,17)

InChI key

NWBJYWHLCVSVIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

6-Benzylaminopurine was used as a plant growth regulator. It was also used to establish and screen the optimal rapid propagation technology of S. tonkinensis by orthogonal test.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shenggang Pan et al.
Rice (New York, N.Y.), 6(1), 9-9 (2013-11-28)
Plant growth regulators play important roles in plant growth and development, but little is known about roles of plant growth regulators in yield, grain qualities and antioxidant enzyme activities in super hybrid rice. In this study, gibberellic acid(GA3), paclobutrazol (PBZ)
Wei Kun-Hua et al.
Pharmacognosy magazine, 9(36), 323-330 (2013-10-15)
Sophora tonkinensis Gapnep. is an important rare medicinal plant in China. There were only a few papers on the rapid propagation of S. tonkinensis through in vitro tissue culture, and still no report focuses on the quality analysis of in
Feng Xu et al.
Journal of agricultural and food chemistry, 60(1), 234-240 (2011-12-14)
The effect of 6-benzylaminopurine (6-BA) on the color, antioxidant activity, and contents of total phenols, glucosinolate, and sulforaphane in broccoli florets was investigated. The results showed that 6-BA treatment markedly inhibited the increase of the L* value and malondialdehyde (MDA)
Michael A Djordjevic et al.
Journal of experimental botany, 62(13), 4649-4659 (2011-06-03)
CLE (CLAVATA3/ESR-related) peptides are developmental regulators that are secreted into the apoplast. Little is known about the role of the sequences that flank CLE peptides in terms of their biological activity or how they are targeted by proteases that are
Nils Braun et al.
Plant physiology, 158(1), 225-238 (2011-11-03)
The function of PsBRC1, the pea (Pisum sativum) homolog of the maize (Zea mays) TEOSINTE BRANCHED1 and the Arabidopsis (Arabidopsis thaliana) BRANCHED1 (AtBRC1) genes, was investigated. The pea Psbrc1 mutant displays an increased shoot-branching phenotype, is able to synthesize strigolactone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service