Skip to Content
Merck
All Photos(2)

Key Documents

129593

Sigma-Aldrich

Malonamide

97%

Synonym(s):

Malonodiamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(CONH2)2
CAS Number:
Molecular Weight:
102.09
Beilstein:
1751401
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

172-175 °C (lit.)

fluorescence

λex 367 nm; λem 445 nm (α-keto acid adduct)

SMILES string

NC(=O)CC(N)=O

InChI

1S/C3H6N2O2/c4-2(6)1-3(5)7/h1H2,(H2,4,6)(H2,5,7)

InChI key

WRIRWRKPLXCTFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The malonamide derivatives are obtained by the one-pot, five-component condensation reaction of isocyanide, Meldrum′s acid, arylidene malononitrile, and two amine molecules in CH2Cl2.

Application

The malonamide-based ionic liquid extractant was used in the extraction of europium(iii) and other trivalent rare-earth ions from nitric acid medium.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana G Neo et al.
Molecular diversity, 15(2), 529-539 (2010-09-03)
A general synthesis of 1,3-dicarbonylic compounds using multicomponent reactions of isocyanides is described. The process involves a Passerini three-component condensation of glyoxal derivatives, isocyanides and acetic acid, followed by metal mediated reductive or solvolytic removal of the acid component. Noteworthy
M M Schiavoni et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 56A(8), 1533-1541 (2000-07-25)
The conformational and tautomeric compositions of malonamide, NH2-C(O)-CH2-C(O)-NH2 were determined by vibrational spectroscopy and theoretical calculations (HF/6-31G*, B3PW91/6-31G*). Solid state Fourier transform infrared and Raman spectra were analysed. They reveal the existence of a diketo tautomer. Theoretical calculations predict a
M J Barlow et al.
Solid state nuclear magnetic resonance, 1(4), 197-204 (1992-11-01)
Methyl tunnel frequencies, measured at 4 K, are found to be 455 +/- 8 kHz in methyl malonamide and 496 +/- 8 kHz in methyl ethyl ketone. The first is unaffected by deuteration of the amide groups. Measurements of the
L C Cerny et al.
Critical care medicine, 11(9), 739-743 (1983-09-01)
This study examines some of the biophysical properties of mixtures of whole blood with 3 different hydroxyethyl starch-hemoglobin (HES-Hgb) polymer blood substitutes. The polymers have about a 3-fold range of number average molecular weight and intrinsic viscosity range. The range
Romain Diss et al.
Physical chemistry chemical physics : PCCP, 7(2), 264-272 (2005-01-21)
According to molecular dynamics simulations, uncomplexed malonamide ligands L and their neutral Eu(NO3)3L2 or charged EuL4(3+) complexes are surface active and adsorb at a water-"oil" interface, where "oil" is modeled by chloroform. Aqueous solvation at the interface is found to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service