Skip to Content
Merck
All Photos(2)

Documents

108421

Sigma-Aldrich

Methyl cyanoacetate

99%

Synonym(s):

Methyl 2-cyanoacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCH2COOCH3
CAS Number:
Molecular Weight:
99.09
Beilstein:
773945
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39031513
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.41 (vs air)

Quality Level

vapor pressure

0.2 mmHg ( 20 °C)

Assay

99%

refractive index

n20/D 1.417 (lit.)

bp

204-207 °C (lit.)

mp

−13 °C (lit.)

density

1.123 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CC#N

InChI

1S/C4H5NO2/c1-7-4(6)2-3-5/h2H2,1H3

InChI key

ANGDWNBGPBMQHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl cyanoacetate is the intermediate product in pharmaceutical organic synthesis as well as in the synthesis of some biologically active compounds used in agriculture. It undergoes calcite or fluorite catalyzed Knövenagel condensation with aromatic aldehydes, giving the corresponding arylidenemalononitriles and (E)-α-cyanocinnamic esters.

Application

Methyl cyanoacetate may be used in the synthesis of various 1,2,5-tricarbonyl compounds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Conformational isomerism in methyl cyanoacetate: A combined matrix-isolation infrared spectroscopy and molecular orbital study.
Reva ID, et al
Physical Chemistry Chemical Physics, 3(19), 4235-4241 (2011)
Construction of 1, 2, 5-Tricarbonyl Compounds using Methyl Cyanoacetate as a Glyoxylate Anion Synthon Combined with Copper (I) Iodide-Catalyzed Aerobic Oxidation.
Kim SH, et al.
Advanced Synthesis & Catalysis, 353(18), 3335-3339 (2011)
Calcite and fluorite as catalyst for the Knovenagel condensation of malononitrile and methyl cyanoacetate under solvent-free conditions.
Wada S and Suzuki H.
Tetrahedron Letters, 44(2), 399-401 (2003)
Søren S Donau et al.
Chemosphere, 233, 873-878 (2019-07-26)
The cyanoacrylate ethyl (2Z)-3-amino-2-cyano-3-phenylacrylate (phenamacril), has been introduced as an effective agent against several fungi species belonging to the Fusarium genus. However, in current literature, knowledge about the environmental behavior of this fungicide is limited and there are no data
Yulia Kaluzhny et al.
Alternatives to laboratory animals : ATLA, 43(2), 101-127 (2015-05-23)
The 7th Amendment to the EU Cosmetics Directive and the EU REACH Regulation have reinforced the need for in vitro ocular test methods. Validated in vitro ocular toxicity tests that can predict the human response to chemicals, cosmetics and other

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service