Skip to Content
Merck
All Photos(2)

Documents

127914

Sigma-Aldrich

1-Phenyl-3-pyrazolidinone

97%

Synonym(s):

Phenidone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10N2O
CAS Number:
Molecular Weight:
162.19
Beilstein:
131856
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

119-121 °C (lit.)

SMILES string

O=C1CCN(N1)c2ccccc2

InChI

1S/C9H10N2O/c12-9-6-7-11(10-9)8-4-2-1-3-5-8/h1-5H,6-7H2,(H,10,12)

InChI key

CMCWWLVWPDLCRM-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

1-Phenyl-3-pyrazolidinone was used as a photograph developing agent.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

275.5 °F - Pensky-Martens closed cup

Flash Point(C)

135.3 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Oxidation of 1-Phenyl-4, 4-dimethyl-3-pyrazolidinone in Alkaline Solution.
The Journal of Organic Chemistry, 30(8), 2571-2575 (1965)
H C Kim et al.
Brain research, 874(1), 15-23 (2000-08-11)
Acculmulating evidence indicates that a marked generation of oxygen free radicals derived from the metabolism of arachidonic acid causes neurodegeneration. Recently, we have demonstrated that the novel antioxidant actions mediated by phenidone, a dual inhibitor of cyclooxygenase/lipoxygenase pathways, may play
Hasan Tunaz
Archives of insect biochemistry and physiology, 63(3), 93-100 (2006-10-19)
Influence of fungal species (conidia spores) on mortality of Pieris brassicae larvae differed when injected into the larvae. The effects of B. bassiana (ARSEF-1151) were expressed in a conidial dose-dependent manner on mortality of the larvae. An increased and faster
A Wakatsuki et al.
Biology of the neonate, 76(2), 84-91 (1999-07-08)
To determine whether ischemia followed by subsequent reperfusion can induce fetal cerebral oxidative damage, we created a model of fetal ischemia/reperfusion using rats at day 19 of pregnancy. Fetal ischemia was induced by unilateral occlusion of the utero-ovarian artery for
J Peng et al.
Plant biology (Stuttgart, Germany), 13(2), 276-284 (2011-02-12)
When attacked by herbivores, plants release herbivore-induced plant volatiles (HIPV) that may function in direct defence by repelling herbivores or reducing their growth. Emission of HIPV may also contribute to indirect defence by attracting natural enemies of the herbivore. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service