Skip to Content
Merck
All Photos(3)

Key Documents

125938

Sigma-Aldrich

Dibutyl phosphite

96%

Synonym(s):

Butyl phosphite, Butyl phosphonate, Di-n-butyl hydrogen phosphite, Di-n-butyl phosphite, Dibutoxyphosphine oxide, Dibutyl hydrogen phosphite, Dibutyl phosphonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2O)2P(O)H
CAS Number:
Molecular Weight:
194.21
Beilstein:
1099706
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39110212
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

96%

impurities

≤1% butanol

refractive index

n20/D 1.423 (lit.)

bp

118-119 °C/11 mmHg (lit.)

solubility

H2O: soluble 7.3 g/L at 25 °C

density

0.995 g/mL at 25 °C (lit.)

SMILES string

CCCCO[PH](=O)OCCCC

InChI

1S/C8H19O3P/c1-3-5-7-10-12(9)11-8-6-4-2/h12H,3-8H2,1-2H3

InChI key

NFJPGAKRJKLOJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibutyl phosphite undergoes addition reaction with Schiff bases to yield 1,4-bis[N-methyl(dibutoxyphosphonyl)-1-(2-furyl)]diaminobenzene and 4,4′-bis[N-methyl(dibutoxyphosponyl)-1-phenyl]benzidine. It is antiwear and extreme-pressure additive in industrial gear oils.

Application

Dibutyl phosphite was used as anti-wear agent in bio-based lubricating oil.

Other Notes

Contains tributyl phosphate

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

244.4 °F - closed cup

Flash Point(C)

118 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Industrial gear oil-a study of the interaction of antiwear and extreme-pressure additives.
Fang X, et al.
Tribology International, 26(6), 395-398 (1993)
Addition products of dibutyl phosphite to schiff bases.
Kraicheva I.
Phosph. Sulfur Relat. Elem., 134(1), 287-294 (1998)
Bio-Lubricants Development: The Potential Use of Boron-Containing Additives.
Dicky D and Dyah SP.
Proceeding of International Conference on Chemical and Material Engineering (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service