Skip to Content
Merck
All Photos(2)

Key Documents

122793

Sigma-Aldrich

Allyl cyanide

98%

Synonym(s):

3-Butenenitrile, Vinylacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2CN
CAS Number:
Molecular Weight:
67.09
Beilstein:
605352
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

autoignition temp.

851 °F

refractive index

n20/D 1.405 (lit.)

bp

116-121 °C (lit.)

density

0.834 g/mL at 25 °C (lit.)

SMILES string

C=CCC#N

InChI

1S/C4H5N/c1-2-3-4-5/h2H,1,3H2

InChI key

SJNALLRHIVGIBI-UHFFFAOYSA-N

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Ryo Yazaki et al.
Journal of the American Chemical Society, 130(44), 14477-14479 (2008-10-11)
Direct catalytic asymmetric addition of allylic cyanides to N-diphenylphosphinoyl ketoimines with a bimetallic catalytic system comprising Ph-BPE/[Cu(CH3CN)4]ClO4/LiOAr is described. Intermediary alpha-adducts readily isomerized to afford synthetically useful alpha,beta-unsaturated nitriles bearing an optically active tetrasubstituted carbon. Applicability to aromatic, heteroaromatic, and
Katja Witzel et al.
PloS one, 8(9), e71877-e71877 (2013-09-17)
The soil-borne fungal pathogen Verticillium longisporum is able to penetrate the root of a number of plant species and spread systemically via the xylem. Fumigation of Verticillium contaminated soil with Brassica green manure is used as an environmentally friendly method
Asymmetric synthesis of isothiazoles through Cu catalysis: direct catalytic asymmetric conjugate addition of allyl cyanide to α,β-unsaturated thioamides.
Yuka Yanagida et al.
Angewandte Chemie (International ed. in English), 50(34), 7910-7914 (2011-07-07)
Marja Tolonen et al.
Journal of agricultural and food chemistry, 50(23), 6798-6803 (2002-10-31)
The formation of plant-derived biomolecules during sauerkraut fermentation was studied. Cabbage was fermented with a starter culture, and the results were compared to the results of spontaneous fermentation. The concentration of flavonoids and glucosinolates was analyzed by HPLC, and that
Ryo Yazaki et al.
Journal of the American Chemical Society, 132(15), 5522-5531 (2010-03-27)
We report that a hard Lewis base substantially affects the reaction efficiency of direct catalytic asymmetric gamma-addition of allyl cyanide (1a) to ketones promoted by a soft Lewis acid/hard Brønsted base catalyst. Mechanistic studies have revealed that Cu/(R,R)-Ph-BPE and Li(OC(6)H(4)-p-OMe)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service