122505
2,3-Dihydroxypyridine
95%
Synonym(s):
2,3-Pyridinediol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H5NO2
CAS Number:
Molecular Weight:
111.10
Beilstein:
109848
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
mp
245 °C (dec.) (lit.)
SMILES string
Oc1cccnc1O
InChI
1S/C5H5NO2/c7-4-2-1-3-6-5(4)8/h1-3,7H,(H,6,8)
InChI key
GGOZGYRTNQBSSA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
2,3-Dihydroxypyridine(DHP) was used in the synthesis of DHP loaded amberlite XAD-2 via azo coupler. It was used in the synthesis of new macromolecular chelators by loading DHP on cellulose via linkers -NH-CH2-CH2-NH-SO2-C6H4-N=N- and -SO2-C6H4-N=N.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Lei Wang et al.
European journal of medicinal chemistry, 156, 680-691 (2018-07-23)
Human immunodeficiency virus (HIV) reverse transcriptase (RT)-associated ribonuclease H (RNase H) remains an unvalidated drug target. Reported HIV RNase H inhibitors generally lack significant antiviral activity. We report herein the design, synthesis, biochemical and antiviral evaluations of a new 6-biphenylmethyl
Jayakanth Kankanala et al.
European journal of medicinal chemistry, 141, 149-161 (2017-10-17)
Human immunodeficiency virus (HIV) reverse transcriptase (RT) associated ribonuclease H (RNase H) is the only HIV enzymatic function not targeted by current antiviral drugs. Although various chemotypes have been reported to inhibit HIV RNase H, few have shown significant antiviral
2, 3-Dihydroxypyridine loaded amberlite XAD-2 (AXAD-2-DHP): preparation, sorption-desorption equilibria with metal ions, and applications in quantitative metal ion enrichment from water, milk and vitamin samples.
Venkatesh G, et al.
Microchimica Acta, 149(3-4), 213-221 (2005)
A al-Dehneh et al.
Journal of animal science, 72(2), 415-420 (1994-02-01)
Sixteen Alpine wethers (average BW 35 +/- 2 kg) were used to evaluate the effect of continuous 48-h intravenous infusions of saline (CON), mimosine (MIM; 200 mg.kg.75.d-1), 2-hydroxy-3(1H)-pyridine (2,3-DHP; 200 mg.kg.75.d-1, or MIM+2,3-DHP (100 mg of MIM plus 100 mg
The first mu(6)-peroxide transition-metal complex: [Ni(8)(L)(12)(O(2))](2+).
Emma J Brown et al.
Angewandte Chemie (International ed. in English), 44(9), 1392-1395 (2005-01-05)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service