Skip to Content
Merck
All Photos(1)

Documents

11030

Sigma-Aldrich

L-Arginine methyl ester dihydrochloride

≥98.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN=C(NH2)NH(CH2)3CH(NH2)COOCH3 · 2HCl
CAS Number:
Molecular Weight:
261.15
Beilstein:
4159929
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (AT)

form

powder

optical activity

[α]20/D +21±1°, c = 2.5% in methanol

reaction suitability

reaction type: solution phase peptide synthesis

color

white

mp

~190 °C (dec.)

application(s)

peptide synthesis

SMILES string

Cl.Cl.COC(=O)[C@@H](N)CCCNC(N)=N

InChI

1S/C7H16N4O2.2ClH/c1-13-6(12)5(8)3-2-4-11-7(9)10;;/h5H,2-4,8H2,1H3,(H4,9,10,11);2*1H/t5-;;/m0../s1

InChI key

XQYZOBNLCUAXLF-XRIGFGBMSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alaleh Asghari-Roodsari et al.
Reproductive sciences (Thousand Oaks, Calif.), 17(4), 391-400 (2010-03-17)
In this study, we explained that exogenous cannabinoid, Delta(9)-tetrahydrocannabinol (THC), has a preventive effect in a murine model of lipopolysaccharide (LPS)-induced preterm delivery and the contribution of nitric oxide (NO) pathway as a mechanism involved in this process. Preterm delivery
J A Simonsen et al.
Acta physiologica (Oxford, England), 198(2), 125-132 (2009-09-30)
We wanted to stabilize the availability of nitric oxide (NO) at levels compatible with normal systemic haemodynamics to provide a model for studies of complex regulations in the absence of changes in NO levels. Normal volunteers (23-28 years) were infused
Xiao-jun Zhang et al.
Clinical nutrition (Edinburgh, Scotland), 27(4), 649-656 (2008-03-05)
L-arginine may play a role in regulation of protein metabolism. The aim of the present study was to investigate the effect of L-arginine supplementation on protein metabolism in skin wound and muscle in anesthetized rabbits. L-[ring-(13)C(6)]phenylalanine was infused as a
Farah Nurlidar et al.
Journal of biomaterials science. Polymer edition, 28(14), 1480-1496 (2017-05-19)
Polyion complex (PIC) gel of poly(Pro-Hyp-Gly) was successfully fabricated by simply mixing polyanion and polycation derivatives of poly(Pro-Hyp-Gly), a collagen-like polypeptide. The polyanion, succinylated poly(Pro-Hyp-Gly), and the polycation, arginylated poly(Pro-Hyp-Gly), contain carboxy (pK
Maria Teresa Calejo et al.
International journal of pharmaceutics, 436(1-2), 454-462 (2012-07-24)
Ethyl(hydroxyethyl) cellulose (EHEC) is known to form hydrogels in water at elevated temperatures in the presence of an ionic surfactant. In this paper, the potential use of arginine-based surfactants is explored considering the production of a low toxicity thermoresponsive hydrogel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service