Skip to Content
Merck
All Photos(1)

Documents

A2883

Sigma-Aldrich

L-Arginine ethyl ester dihydrochloride

≥98%

Synonym(s):

L-Arginine ethyl ester hydrochloride (1:2)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N4O2 · 2HCl
CAS Number:
Molecular Weight:
275.18
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

L-Arginine ethyl ester dihydrochloride,

Assay

≥98%

Quality Level

form

powder

color

white

storage temp.

−20°C

SMILES string

Cl.CCOC(=O)C(N)CCCNC(N)=N

InChI

1S/C8H18N4O2.ClH/c1-2-14-7(13)6(9)4-3-5-12-8(10)11;/h6H,2-5,9H2,1H3,(H4,10,11,12);1H

InChI key

DUCUWLZKZPQJDY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soyoung Shin et al.
Biochemical and biophysical research communications, 414(4), 660-663 (2011-10-12)
We examined the relative contributory roles of extracellular vs. intracellular L-arginine (ARG) toward cellular activation of endothelial nitric oxide synthase (eNOS) in human endothelial cells. EA.hy926 human endothelial cells were incubated with different concentrations of (15)N(4)-ARG, ARG, or L-arginine ethyl
N E Paul et al.
Bio-medical materials and engineering, 25(3), 289-297 (2015-09-26)
Adipose tissue engineering is a promising solution for the reconstruction of soft tissue defects. An insufficient neovascularisation within the scaffolds that leads to necrosis and tissue loss is still a major shortcoming of current tissue engineering attempts. Biomaterials, which release

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service