Skip to Content
Merck
All Photos(1)

Key Documents

10849

Sigma-Aldrich

Mowiol® 28-99

Mw ~145,000

Synonym(s):

Poly(vinyl alcohol)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.23

mol wt

Mw ~145,000

extent of labeling

99.0-99.8 mol% hydrolysis
~3,300 polymerization

ign. residue

≤0.5%

viscosity

26-30 mPa.s, 4 % in H2O(20 °C)

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Mowiol is a registered trademark of Kuraray Europe GmbH

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT SE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J H Seol et al.
Rhinology, 51(2), 137-142 (2013-05-15)
A number of different nasal packing materials are available for prevention of nasal bleeding after endoscopic sinus surgery. Polyvinyl alcohol (PVA) coated nasal packing is an expandable packing for use in rhinological surgery. This innovative surf- ace treatment helps to
Miao Zhang et al.
Journal of visualized experiments : JoVE, (73)(73), e4335-e4335 (2013-03-15)
Existing protocols for the generation of neurons from human pluripotent stem cells (hPSCs) are often tedious in that they are multistep procedures involving the isolation and expansion of neural precursor cells, prior to terminal differentiation. In comparison to these time-consuming
Cijun Shuai et al.
Biofabrication, 5(1), 015014-015014 (2013-02-07)
A tetragonal polyvinyl alcohol (PVA) scaffold with 3D orthogonal periodic porous architecture was fabricated via selective laser sintering (SLS) technology. The scaffold was fabricated under the laser power of 8 W, scan speed of 600 mm min(-1), laser spot diameter of 0.8 mm
Sreejith Raveendran et al.
Carbohydrate polymers, 92(2), 1225-1233 (2013-02-13)
Extremophilic bacterial polysaccharide based biocompatible nanofibers were produced for the first time via electrospinning technique. Mauran (MR), an extremophilic sulfated exopolysaccharide was extracted from moderately halophilic bacterium, Halomonas maura and characterized for the application of nanofiber synthesis. Thin-uniform MR nanofibers
Feng-yong Liu et al.
Acta radiologica (Stockholm, Sweden : 1987), 54(1), 61-66 (2013-02-05)
Preoperative embolization of tumors is a well-established procedure that has been successfully applied in various clinical situations. Preoperative embolization can reduce the vascularity of tumors resulting in a clearer operative field, less difficult dissection, decreased blood loss, and, in some

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service