Skip to Content
Merck
All Photos(2)

Key Documents

105937

Sigma-Aldrich

3,4,4′-Trichlorocarbanilide

99%

Synonym(s):

1-(4-Chlorophenyl)-3-(3,4-dichlorophenyl)urea, TCC, Triclocarban

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NHCONHC6H4Cl
CAS Number:
Molecular Weight:
315.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Agency

EPA 1694

Quality Level

vapor pressure

<0.1 mmHg ( 25 °C)

Assay

99%

form

solid

mp

254-256 °C (lit.)

solubility

methanol: soluble

application(s)

environmental

functional group

amine
chloro

SMILES string

Clc1ccc(NC(=O)Nc2ccc(Cl)c(Cl)c2)cc1

InChI

1S/C13H9Cl3N2O/c14-8-1-3-9(4-2-8)17-13(19)18-10-5-6-11(15)12(16)7-10/h1-7H,(H2,17,18,19)

InChI key

ICUTUKXCWQYESQ-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

General description

3,4,4′-Trichlorocarbanilide(TCC) is employed as bacteriostat and antimicrobial agent in bar soaps, cosmetics and other personal care prducts.TCC is a toxic, persistent and potentially bioaccumulative polychlorinated binuclear aromatic urea pesticide.

Application

3,4,4′-Trichlorocarbanilide was used to develop a sensitive analytical method to determine triclocarban and environmental phenols in breast milk using isotope dilution on-line solid-phase extraction coupled with HPLC–tandem mass spectrometry. 3,4,4′-Trichlorocarbanilide was used in quantitative structure-activity relationship (QSAR)analyses conducted to study the co-occurrence of Triclocarban and Triclosan in U.S. Water Resources.

Biochem/physiol Actions

3,4,4′-Trichlorocarbanilide interferes with mammalian reproduction and can cause methemoglobinemia in humans.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biodegradation of 3, 4, 4'-trichlorocarbanilide, TCC?, in sewage and activated sludge.
Gledhill WE.
Water Research, 9(7), 649-654 (1975)
Rong Chu et al.
Oncotarget, 7(48), 78958-78970 (2016-10-16)
Microtubule targeting agents (MTAs) characteristically promote phosphorylation and degradation of Mcl-1, and this represents a critical pro-apoptotic signal in mitotic death. While several phosphorylation sites and kinases have been implicated in mitotic arrest-induced Mcl-1 phosphorylation, a comprehensive biochemical analysis has
Rolf U Halden et al.
Environmental science & technology, 39(6), 1420-1426 (2005-04-12)
Triclocarban (TCC) and triclosan (TCS) are antimicrobial additives in personal care products. Whereas TCS has been studied extensively, the environmental fate of TCC remains largely unknown. To address this data gap, we performed quantitative structure-activity relationship (QSAR) analyses that suggested
Arjun K Venkatesan et al.
Journal of hazardous materials, 229-230, 29-35 (2012-06-30)
The antimicrobial agents triclosan (TCS), triclocarban (TCC) and their associated transformation products are of increasing concern as environmental pollutants due to their potential adverse effects on humans and wildlife, including bioaccumulation and endocrine-disrupting activity. Analysis by tandem mass spectrometry of
Jian-Liang Zhao et al.
Water research, 47(1), 395-405 (2012-11-07)
Triclosan (TCS) and triclocarban (TCC) are two commonly used personal care products. They may enter into aquatic environments after consumption and pose potential risks to aquatic organisms. We investigated the occurrence and fate of TCS and TCC in five large

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service