Merck
  • Intratumoral gold-doxorubicin is effective in treating melanoma in mice.
Intratumoral gold-doxorubicin is effective in treating melanoma in mice.
Nanomedicine : nanotechnology, biology, and medicine (2015-04-19)
Xuan Zhang, Jose G Teodoro, Jay L Nadeau
ABSTRACT
Intratumoral injection of ultra-small gold nanoparticles (AuNPs) conjugated to doxorubicin (Au-Dox) is effective against both murine B16 and human SK-MEL-28 tumors in mice. Au-Dox suppresses growth of B16 tumors in immunocompetent mice by >70% for at least 19 days. In SK-MEL-28 xenografts, Au-Dox suppresses tumor growth almost completely for >13 weeks, while tumors treated with Dox alone demonstrate accelerated growth after 10 weeks. Histological analysis shows significant apoptosis and necrosis in Au-Dox treated tumors. Intratumoral injection is significantly more effective than intravenous injection, which leads to significant accumulation in liver and kidney with sub-therapeutic concentrations of Au-Dox. However, IV injection does not lead to significant damage in non-target organs, so improved targeting should permit this mode of delivery with little risk of systemic toxicity. The current construct is suitable for tumors accessible to intratumoral injection and represents a viable approach doxorubicin-resistant solid tumors. Drug resistance is a significant problem in the fight against cancer. The authors describe a new approach in combating drug resistance in tumor cells by conjugating ultrasmall gold nanoparticles to doxorubicin. They tested the efficacy in in-vivo models using two melanoma cell lines. The promising results obtained from intra-tumoral injections contribute a way in future drug designs showing that conjugation to nanoparticles could lead to more effective and synergistic killing of tumor cells.
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 0.1 M in ethanol
Sigma-Aldrich
Hydrochloric acid solution, 12 M
Sigma-Aldrich
Hydrochloric acid solution, 2 M
Sigma-Aldrich
Hydrochloric acid solution, 1 M
Sigma-Aldrich
Hydrochloric acid solution, 0.5 M
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 0.2 M
Sigma-Aldrich
Hydrochloric acid solution, 0.05 M
Sigma-Aldrich
Hydrochloric acid solution, 0.02 M
Sigma-Aldrich
Hydrochloric acid solution, 0.01 M
Sigma-Aldrich
Hydrochloric acid solution, 6 M
Sigma-Aldrich
Hydrochloric acid, JIS special grade, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid, SAJ first grade, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%