Skip to Content
Merck
All Photos(1)

Key Documents

13-1710

Sigma-Aldrich

Hydrochloric acid

0.5 M

Synonym(s):

Hydrogen chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:

Product Name

Hydrochloric acid solution, 0.5 M

grade

reagent

vapor pressure

190 mmHg ( 25 °C)

form

liquid

availability

available only in Japan

concentration

0.5 M
1/2 N

bp

100 °C (lit.)

density

1.01 g/cm3 at 20 °C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siofra Christine Harrington et al.
The British journal of ophthalmology, 103(8), 1112-1118 (2018-10-14)
To report refractive error prevalence and visual impairment in Republic of Ireland (henceforth 'Ireland') schoolchildren. The Ireland Eye Study examined 1626 participants (881 boys, 745 girls) in two age groups, 6-7 years (728) and 12-13 years (898), in Ireland between
Charlotte Strandgren et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(8), 3193-3205 (2015-04-17)
Hutchinson-Gilford progeria syndrome (HGPS) is a rare premature aging disorder that is most commonly caused by a de novo point mutation in exon 11 of the LMNA gene, c.1824C>T, which results in an increased production of a truncated form of
Brian W Simons et al.
The Journal of pathology, 235(3), 478-489 (2014-10-29)
Inflammation is associated with several diseases of the prostate including benign enlargement and cancer, but a causal relationship has not been established. Our objective was to characterize the prostate inflammatory microenvironment after infection with a human prostate-derived bacterial strain and
Aparajita H Chourasia et al.
EMBO reports, 16(9), 1145-1163 (2015-08-02)
BNip3 is a hypoxia-inducible protein that targets mitochondria for autophagosomal degradation. We report a novel tumor suppressor role for BNip3 in a clinically relevant mouse model of mammary tumorigenesis. BNip3 delays primary mammary tumor growth and progression by preventing the
Shu Zhang et al.
PloS one, 10(7), e0131833-e0131833 (2015-07-07)
Cyclin-dependent kinase 5 (CDK5) is a cytoplasmic serine/ threonine kinase. Knockdown of CDK5 enhances paclitaxel sensitivity in human ovarian cancer cells. This study explores the mechanisms by which CDK5 regulates paclitaxel sensitivity in human ovarian cancers. Multiple ovarian cancer cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service