Skip to Content
Merck
All Photos(1)

Key Documents

320331

Sigma-Aldrich

Hydrochloric acid

37%, ACS reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
Beilstein:
1098214
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.21

Product Name

Hydrochloric acid, ACS reagent, 37%

grade

ACS reagent

Quality Level

Agency

suitable for GB 5009.253-2016
suitable for SM 5210

vapor density

1.3 (vs air)

vapor pressure

3.23 psi ( 21.1 °C)
7.93 psi ( 37.7 °C)

Assay

36.5-38.0% (ACS specification)
37%

form

liquid

impurities

≤1 ppm free chlorine

ign. residue

≤5 ppm

color

APHA: ≤10, free from suspended matter or sediment

bp

>100 °C (lit.)

density

1.18 g/L at 20 °C

anion traces

bromide (Br-): ≤0.005%
sulfate (SO42-): ≤1 ppm
sulfite (SO32-): ≤1 ppm

cation traces

As: ≤0.01 ppm
Fe: ≤0.2 ppm
NH4+: ≤3 ppm
heavy metals: ≤1 ppm (by ICP)

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrochloric acid (Reagent grade) is suitable for general lab applications and tests with low specification controls.,
Applications include:      
  • General Lab Use      
  • General Chemical Synthesis      
  • Purification      
  • Cleaning

Application


  • Unlocking high-performance HCl adsorption at elevated temperatures: This research demonstrates the use of hydrochloric acid solution for synthesizing robust Ca-Mg-Al mixed oxides. These materials are crucial in environmental applications for adsorbing acidic gases, showcasing hydrochloric acid′s role in developing solutions to combat air pollution (Cao et al., 2024).

  • Sustainable and Green Corrosion Inhibition of Mild Steel: Hydrochloric acid is utilized here to test new, environmentally friendly corrosion inhibitors. This is vital in industries where maintaining material integrity against chemical attack is crucial, highlighting hydrochloric acid′s role in innovative material science applications (Rizi et al., 2023).

  • Synthesis of Zinc Oxide Nanorods from Zinc Borate Precursor: This study uses hydrochloric acid in the synthesis of zinc oxide nanorods, important for supercapacitor applications. It underscores the critical role of hydrochloric acid in nanomaterials research, pushing the boundaries of energy storage technology (Lefdhil et al., 2023).

  • An improved digestion and analysis procedure for silicon in plant tissue: Hydrochloric acid is employed to digest plant tissues for silicon content analysis, enhancing agricultural and biological research methodologies. This application reflects the importance of hydrochloric acid in scientific studies that require precise chemical processing (Langenfeld and Bugbee, 2023).

  • N-N(+) Bond-Forming Intramolecular Cyclization of O-Tosyloxy β-Aminopropioamidoximes and Ion Exchange Reaction: In this research, hydrochloric acid plays a key role in organic synthesis, demonstrating its versatility in pharmaceutical compound development. The study provides insights into the synthesis of complex organic compounds, which are vital in medicinal chemistry (Kayukova et al., 2023).

Packaging

Single 2.5L units require a can opener to open the metal shipping container.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clay Swackhamer et al.
Food & function, 10(8), 5174-5187 (2019-08-06)
Previous studies have shown that the size of almond particles influences lipid bioaccessibility during digestion. However, the extent of structural breakdown of almond particles during gastric digestion and its impact on lipid bioaccessibility is unclear. In this study, in vitro
Bilal Çakir et al.
Plant foods for human nutrition (Dordrecht, Netherlands), 74(3), 414-420 (2019-07-07)
The amount of cold press oil manufacture is globally rising, which in turn leads to the accumulation of deoiled plant seeds at significant quantities and consequent manufacture of plant protein products. In this study, we made an attempt to analyze
Xuan Zhang et al.
Nanomedicine : nanotechnology, biology, and medicine, 11(6), 1365-1375 (2015-04-19)
Intratumoral injection of ultra-small gold nanoparticles (AuNPs) conjugated to doxorubicin (Au-Dox) is effective against both murine B16 and human SK-MEL-28 tumors in mice. Au-Dox suppresses growth of B16 tumors in immunocompetent mice by >70% for at least 19 days. In
Jay G Hosking et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(4), 1005-1015 (2014-10-21)
Successful decision making often requires weighing a given option's costs against its associated benefits, an ability that appears perturbed in virtually every severe mental illness. Animal models of such cost/benefit decision making overwhelmingly implicate mesolimbic dopamine in our willingness to
Mahsa Sadeghi et al.
British journal of pharmacology, 172(2), 460-468 (2014-01-01)
Tapentadol is a novel analgesic that combines moderate μ-opioid receptor agonism and noradrenaline reuptake inhibition in a single molecule. Both mechanisms of action are involved in producing analgesia; however, the potency and efficacy of tapentadol in individual neurons has not

Articles

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service