Merck
  • Ceftriaxone attenuates acute cocaine-evoked dopaminergic neurotransmission in the nucleus accumbens of the rat.
Ceftriaxone attenuates acute cocaine-evoked dopaminergic neurotransmission in the nucleus accumbens of the rat.
British journal of pharmacology (2015-09-17)
J L Barr, B A Rasmussen, C S Tallarida, J L Scholl, G L Forster, E M Unterwald, S M Rawls
ABSTRACT
Ceftriaxone is a β-lactam antibiotic and glutamate transporter activator that reduces the reinforcing effects of psychostimulants. Ceftriaxone also reduces locomotor activation following acute psychostimulant exposure, suggesting that alterations in dopamine transmission in the nucleus accumbens contribute to its mechanism of action. In the present studies we tested the hypothesis that pretreatment with ceftriaxone disrupts acute cocaine-evoked dopaminergic neurotransmission in the nucleus accumbens. Adult male Sprague-Dawley rats were pretreated with saline or ceftriaxone (200 mg kg(-1) , i.p. × 10 days) and then challenged with cocaine (15 mg kg(-1) , i.p.). Motor activity, dopamine efflux (via in vivo microdialysis) and protein levels of tyrosine hydroxylase (TH), the dopamine transporter and organic cation transporter as well as α-synuclein, Akt and GSK3β were analysed in the nucleus accumbens. Ceftriaxone-pretreated rats challenged with cocaine displayed reduced locomotor activity and accumbal dopamine efflux compared with saline-pretreated controls challenged with cocaine. The reduction in cocaine-evoked dopamine levels was not counteracted by excitatory amino acid transporter 2 blockade in the nucleus accumbens. Pretreatment with ceftriaxone increased Akt/GSK3β signalling in the nucleus accumbens and reduced levels of dopamine transporter, TH and phosphorylated α-synuclein, indicating that ceftriaxone affects numerous proteins involved in dopaminergic transmission. These results are the first evidence that ceftriaxone affects cocaine-evoked dopaminergic transmission, in addition to its well-described effects on glutamate, and suggest that its ability to attenuate cocaine-induced behaviours, such as psychomotor activity, is due in part to reduced dopaminergic neurotransmission in the nucleus accumbens.
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, ≥98%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, BioXtra
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in H2O
Sigma-Aldrich
Trimethylamine solution, 43.0-49.0% in H2O (T)
Sigma-Aldrich
Trimethylamine solution, 31-35 wt. % in ethanol, 4.2 M, contains toluene as stabilizer
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in propylene glycol
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil
Sigma-Aldrich
Sodium hydride, dry, 90%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles