Merck
  • Controlled self-assembly of a pyrene-based bolaamphiphile by acetate ions: from nanodisks to nanofibers by fluorescence enhancement.
Controlled self-assembly of a pyrene-based bolaamphiphile by acetate ions: from nanodisks to nanofibers by fluorescence enhancement.
Soft matter (2015-05-01)
Shouchun Yin, Lin Dong, Yijun Xia, Bin Dong, Xingxing He, Dong Chen, Huayu Qiu, Bo Song
ABSTRACT
In this paper, a pyrene moiety is incorporated into a bolaamphiphile to form a novel molecule denoted PRB. Above the critical micelle concentration, PRB forms nanodisks in the aqueous solution. The addition of acetate ions induces a morphological change in self-assembled aggregates, which convert into nanofibers with a diameter of several nanometers. More interestingly, along with the morphological change, the fluorescence of the assemblies was enhanced concomitantly, which can be attributed to the binding effect of acetate ions on pyridinium head groups of PRB.
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Tetrakis(triphenylphosphine)palladium(0), ≥99.99% trace metals basis
Sigma-Aldrich
Tetrakis(triphenylphosphine)palladium(0), 99%
Sigma-Aldrich
DCC, 1.0 M in methylene chloride
Sigma-Aldrich
3-Butyn-1-ol, 97%
Sigma-Aldrich
DCC, 99%