Merck
  • Effects of polarity, hydrophobicity, and density of ionic liquids on cellulose solubility.
Effects of polarity, hydrophobicity, and density of ionic liquids on cellulose solubility.
Physical chemistry chemical physics : PCCP (2015-11-20)
Mitsuru Abe, Kosuke Kuroda, Daiki Sato, Haruhito Kunimura, Hiroyuki Ohno
ABSTRACT
We have synthesised novel ionic liquids (ILs) to show both cellulose dissolution ability and LCST-type phase transition after mixing with water. To realise both polar and hydrophobic properties, tetraalkylphosphonium cations and a series of carboxylate anions were employed to assume hydrophobic and highly polar properties, respectively. Effects of their alkyl chain length on the water compatibility and cellulose solubility of the corresponding ILs were systematically examined. We succeeded in synthesising novel ILs which dissolve cellulose and separable with water at moderate temperature. Through the present study, we have clarified that not only polarity but also density of ILs is an important factor in designing the ILs for cellulose dissolution.
Sigma-Aldrich
Myristic acid, ≥95%, FCC, FG
Sigma-Aldrich
Myristic acid, natural, ≥98.5%, FG
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
1-Bromopentane, 98%
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Sigma-Aldrich
Decanoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Hexanoic acid, ≥99%
Sigma-Aldrich
Octanoic acid, ≥98%, FG
Sigma-Aldrich
Behenic acid, 99%
Sigma-Aldrich
Octanoic acid, natural, ≥98%, FG
Sigma-Aldrich
Hexanoic acid, ≥98%, FCC, FG
Sigma-Aldrich
Octanoic acid, ≥98%
Sigma-Aldrich
Decanoic acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
1-Bromohexane, 98%
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Myristic acid, Sigma Grade, ≥99%
Sigma-Aldrich
Decanoic acid, ≥98.0%
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Octanoic acid, ≥99%
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Octanoic acid, ≥96.0%
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Sigma-Aldrich
Dodecanoic acid, ≥99% (GC/titration)
Sigma-Aldrich
Dodecanoic acid, 98%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%